Clinical Laboratory Freezers
High performance storage for the lab and clinical environment
Temperature variation can have an impact on the viability and efficacy of vaccines, medication, reagents and other temperature-sensitive materials. These variations can shorten product shelf life, degrade enzymes and reduce vaccine effectiveness. Therefore, it is important to choose a high-performance freezer that has been designed for the demanding standards of the laboratory and clinical environment.
Clinical laboratory freezer categories
The Thermo Scientific™ GPF Series countertop freezer allows for easy transportation and placement within any laboratory or clinical setting, with a countertop footprint about the size of a microwave. These freezers increase user efficiency and lessen physical strain, as there is no bending or squatting required. Top-mounted controls, reversible doors and an ergonomic handle contribute to its modern design, which allows for a convenient, practical approach to a busy day. Multiple GPFs in a medical facility allow for the convenient storage of daily doses of vaccines and pharmaceuticals requiring frozen storage in multiple locations. 
Thermo Scientific™ TSX Series high-performance -30°C auto defrost freezers are designed with features that support sample protection and sustainability objectives for the storage of clinical and laboratory-grade storage requiring -30°C. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Thermo Scientific™ TSX Series high-performance -20°C manual defrost freezers are designed with features that support sample protection and sustainability objectives for the storage of clinical and laboratory-grade storage requiring -20°C. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers are designed with features that support sample protection and sustainability objectives for the storage of enzymes requiring -20°C. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Thermo Scientific™ TSX Series high-performance plasma freezers are designed with features that support sample protection and sustainability objectives for the storage of plasma. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ High-Performance Lab Freezers. Our Revco -30°C and -20°C high-performance freezers are designed for medical and scientific applications such as storage of reagents, pharmaceuticals, biologicals or other commonly used laboratory materials. Models offer cabinet sizes to fit a variety of space needs and storage volume requirements.
Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature digital controls and excellent temperature stability.
Related products
Featured clinical laboratory freezers video
Video Player is loading.
Current Time 0:00
Duration 3:31
Loaded: 0%
Stream Type LIVE
Remaining Time 3:31
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Monitor your cold storage systems
    Put your mind at ease knowing a state-of-the-art system is watching, safeguarding, and data logging critical equipment parameters, and be notified right away if there is a power or mechanical failure.
    Find your wireless solution
    Please note that these products are for research use only. To view our selection of in vitro diagnostic certified instruments, please visit our Diagnostic Testing Instruments and Automation page. 
    *The term "leakproof" applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.