Search Thermo Fisher Scientific
The final part of any genetic analysis assay is the analysis and reporting.We offer a suite of software tools to assist with your laboratory’s goals. Unless otherwise noted, all software tools are for research use only, not for use in diagnostic procedures.
TaqMan® Genotyper™ Software | A free SNP genotyping data analysis tool for use with TaqMan® SNP Genotyping Assays.
| Learn more |
CopyCaller® Software | Free software developed specifically for TaqMan® Copy Number Assay data analysis.
| Learn more |
Variant Reporter® Software | Designed for reference-based and nonreference-based analysis such as mutation detection and analysis, SNP discovery, and sequence confirmation.
| 30 Day Demo |
GeneMapper® Software | GeneMapper is a flexible genotyping software package that provides DNA sizing and quality allele calls for all Applied Biosystems® electrophoresis-based genotyping systems. | Learn more |
LifeScope™ Software | SOLiD® LifeScope™ Software uses a flexible pipeline architecture to provide maximum flexibility and easy of use for performing high-throughput data analysis for SOLiD® instrument data.
| Learn more |
DataAssist™ Software | DataAssist™ Software is a free, easy-to-use data analysis tool that utilizes the comparative Ct (ΔΔCt) method to rapidly and accurately quantitate relative gene expression across a large number of genes and samples. You can import raw data from hundreds of plates or TaqMan® arrays, and change analysis settings including selection of normalization controls and method (single or multiple control genes). It provides instant responses and fast calculations, allows normalization using multiple reference genes, and provides analysis results in content-rich tables and scalable graphic charts that are easily exported. | Learn more |
RealTime StatMiner® Software | Advanced data mining software for real-time PCR data analysis. RealTime StatMiner® Software provides a high-powered, straightforward solution to data analysis for qPCR. | Learn more |
For Research Use Only. Not for use in diagnostic procedures.