Search Thermo Fisher Scientific
Discover our modern, powerful and robust clinical prenatal screening software for trisomies and neural tube defects (NTDs) in the first and second trimester.
Thermo Scientific B·R·A·H·M·S Fast Screen pre I plus Software is a CE marked prenatal screening software designed to ensure convenience of data entry, risk calculation and reporting for laboratories with both low and high data throughput. It is designed to be used with Thermo Scientific B·R·A·H·M·S KRYPTOR Instruments and Assays.
B·R·A·H·M·S Fast Screen pre I plus Software comprises first trimester algorithms for trisomy that are based on the Fetal Medicine Foundation UK (FMF) data and are updated with recently published correction factors. In addition, our software also includes the algorithm from the FMF Germany.
Trisomies 21, 18 and 13 | |
Time range |
|
Biomarkers |
|
Physical markers |
|
View the FMF first trimester combined test performances for fetal trisomies below.3
Risk cut-off | Detection rate | False positive rate | |
Trisomy 21 (Down’s syndrome) |
1:100 | 92.1% | 4.6% |
Trisomy 18 (Edwards' syndrome) |
1:50 | 94.6% | 2.7% |
Trisomy 13 (Patau's syndrome) |
1:50 | 91.1% |
2.7% |
The B·R·A·H·M·S Fast Screen pre I plus second trimester algorithm allows calculation of risk for trisomies 21 and 18 with double test (AFP in combination with free βhCG or total hCG) between 14+0 and 19+6.
Between 15+0 and 19+6, the AFP concentration can be used as a marker of fetal NTD.
Simple and user-friendly interface available in six languages
Available in English, German, French, Spanish, Italian, and Turkish
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
© 2024 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. B·R·A·H·M·S is a registered trademark of B·R·A·H·M·S GmbH.
KRYPTOR is a trademark of Cisbio Bioassays, licensed for use by B·R·A·H·M·S GmbH, a part of Thermo Fisher Scientific. Other product names in this document are used for identification purposes; they may be trademarks and/or registered trademarks of their respective companies.