More rapid, automated, and complete myeloid genomic profiling
Myeloid testing should be fast. However, myeloid malignancy samples can be challenging to analyze. They are complex, heterogeneous, and have the potential to proliferate rapidly. To get meaningful genetic insights, you need a rapid and streamlined approach for studying all key mutations.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Advancements in Rapid NGS for Myeloid and Lymphoid Malignancies: Insights from Dr. Robert Baker
      Join us for an insightful webinar where Dr. Robert Baker will present the transformative impact of rapid next-generation sequencing (NGS) technologies in the field of hematological malignancies. Discover how his laboratory has significantly reduced turnaround times and improved genomic profiling accuracy, streamlining laboratory workflows by integration of the Ion Torrent Genexus System and the Oncomine Myeloid Assay GX v2, which allows for comprehensive profiling and rapid detection of key variants. Don't miss this opportunity to learn about the latest advancements in NGS and their real-world applications.
      Robert Baker
      Molecular Haematology Operations Manager
      Health Services Laboratory (HSL)
      Robert Baker
      The Impact of Fast NGS Molecular Profiling in Myeloid Malignancy Research
      At EHA 2022, Dr. Dolors Colomer discussed the impact that the Oncomine Myeloid Assay GX had on turnaround times and laboratory efficiency. She presented compelling data on FLT3-ITD, showing 100% concordance between NGS and fragment analysis and the ability to detect low-frequency variants ranging in size from 12–180 bp.
      Dolors Colomer, PhD
      Head of Hematopathology
      Hospital Clinic, IDIBAPS,
      University of Barcelona, Spain
      Dolors Colomer
      Oncomine Myeloid Assay GX v2
      The Ion Torrent Oncomine Myeloid Assay GX V2 on the Ion Torrent Genexus System provides a complete next-generation sequencing (NGS) solution that can deliver a comprehensive myeloid mutational report in as little as 24 hours.
      Profile all key myeloid mutations with one test
      Simultaneously interrogate 45 DNA target genes and 34 fusion driver genes (RNA). This broad fusion panel allows you to sequence over 800 unique fusion transcripts. Gene content on the panel has been curated to cover relevant targets for all the major myeloid disorders—AML, MDS, MPN, CML, CMML, and JMML.
      Complete coverage of key genes
      Detect multiple types of mutations at once
      Myeloid samples frequently harbor mutations in difficult-to-sequence regions of the genome. The Oncomine Myeloid Assay GX v2 has been carefully engineered to reliably detect all relevant mutations associated with myeloid malignancies, including targets in challenging genes like CEBPA, FLT3, CALR and more.
      • Single nucleotide variants
      • Insertions and Deletions
      • Tandem duplications
      • Gene fusions
      dna
      View scientific posters
      2024 AACR Poster
      Detection of KMT2A-PTDs in healthy donor and myeloid malignant samples using next generation sequencing
      2024 AACR Poster
      2024 AMP Poster
      Development of an automated genomic profiling assay for myeloid malignancies research
      2024 AMP Poster
      2023 AACR Poster
      Fully automated sample-to-report NGS workflow for comprehensive genomic profiling of myeloid research samples
      2023 AACR Poster
      It's NGS made easy
      A highly integrated workflow lets you go from specimen to report in as little as 20 minutes of hands-on time required. It’s never been easier to implement myeloid NGS testing in your lab.
      Genexus NGS System
      Automate your workflow from specimen to report
      Start with any common myeloid specimen type, including whole blood, bone marrow, or peripheral blood leukocytes. From there, the Genexus System integrates and automates nucleic acid extraction, purification, quantification, library preparation, sequencing, analysis and reporting within a single software ecosystem. 
      blood sample
      Whole Blood
      Peripheral Blood Leukocytes (PBLs)
      Bone Marrow
      extraction
      Nucleic acid extraction and quantification
      Library preparation
      Sequencing
      Reporting & analysis
      report
      Biomarkers linked to relevant evidence from
      public data sources
      Clear and concise reports
      Customizable variant annotation and reporting tools are available. These tools help simplify the bioinformatics workflow and enable you to focus on finding the biological meaning of your data.
      informatics
      Ordering information
      Catalog # Name Size Price (USD) Qty
      A50694 Each
      17,910.00
      A50753 Each
      11,940.00
      A50754 Each
      6,630.00
      Ready to speak to a Thermo Fisher representative?
      We will be happy to answer your questions and provide a demo of our NGS solutions.
      For Research Use Only. Not for use in diagnostic procedures.