The Thermo Scientific Orion3 Electrostatic Discharge Tester is designed to test for potentially destructive effects of ESD, enabling the identification and hardening of protection structures prior to full-scale production. Given the extremely high cost of integrated circuit production, as well as customer demands for precisely timed component delivery schedules, the Orion3 can save substantial costs in terms of time, material and lost opportunity (time-to-market). The high-resolution dual cameras permit easy and rapid discharge pin alignment during the test setup routine. Both the x- and y-axis are displayed simultaneously. The cameras may be left on during the test to monitor the pin alignment. Vacuum hold-down enables device testing regardless of device types and sizes.
The Orion3 is capable of performing FICDM (field induced or air discharge) testing to all popular industry standards. An event detector ensures each discharge occurs as expected, ensuring proper alignment and contact between the device and the system discharge pin. Events are logged and reported upon test completion, providing confirmation of pin discharge, and the indication of non-stressed pins that require retest. Testing can also be performed, using our LI-CCDM contact test method, which both charges and discharges the device through a controlled impedance environment, eliminating testing issues that can be seen when using air discharge methods.
Key Features
Supports all popular test standards
JS-002, JEDEC, ESDA, AEC and LI-CCDM methods.
Ergonomic work process
Bench height test area allows for an ergonomic work process.
High resolution dual cameras
High resolution dual cameras permit easy discharge pin alignment during test plan set-up.
Humidity level control
Environmental monitor within the test area allows control of humidity levels during testing by flooding the area with user supplied dry air or nitrogen.
Event detector
Event detector confirms ESD discharge and reports when a pin is not stressed.
Automatic waveform capture
Automatic waveform capture for calibration or device testing. Stores Ipeak and min/max values for each captured waveform.
Adjustable fixture
Device under test (DUT) held in place by an internal vacuum pump; alignment assured via adjustable fixture.
Thermo Scientific Scimitar software platform
Intuitive test set-up and test execution using the Thermo Scientific Scimitar software platform.
Integrated package outline editor
Integrated package outline editor allows creation of any device, including off grid.
Multiple device definition formats
Device definitions can be imported from legacy applications, other Scimitar applications, and in text based (x, y) formats.
Specifications
Style Sheet for Products Table Specifications
Electrical Requirements
  • 120 or 240 VAC, 15A 50/60Hz
Test Voltage Range
  • ±25 to ±2000 V (±1V steps)
Voltage
  • 120 or 240 VAC, 15A
Description
  • Electrostatic Discharge Tester, Charged device model
Humidity Range
  • 30 to 60% noncondensing (recommended test chamber range is less than 20%)
Item Description
  • Tester; Charged device model
Motion System
  • x,y axis - min. step size 0.001 with 0.00025 in. accuracy (x,y axis - min. step size 25.4µm with 6µm accuracy); z axis - vertical travel to 1.5 with 0.00025 in. accuracy (z axis - vertical travel to 38.1mm with 6µm accuracy)
Specifications
  • Customer Supplied Vacuum: ≥ 16 in. Hg
Details
  • Dry air / nitrogen: 0.25 scfm @ 60 psi max
Temperature Range
  • Operating Temperature: +15° to +30°C (+59° to +86°F); Non-operating temperature: +4° to +60°C (+40° to +140°F)
Hertz
  • 50/60 Hz
Unit Size
  • Each
English
Metric
Test Area
  • 4 x 4 in.
  • 10.16 x 10.16 cm
Size
  • 26D x 23W x 48H in
  • 66D x 59W x 122H cm
Depth 
  • 26 in
  • 66 cm
Height
  • 48 in
  • 122 cm
Width
  • 23 in
  • 59 cm
Weight
  • 300 lb
  • 112 kg
Applications
esd_thumb_274x180_144dpi
ESD Semiconductor Qualification
Every electrostatic discharge (ESD) control plan is required to identify devices that are sensitive to ESD. We offer a complete suite of test systems to help with your device qualification requirements.
Style Sheet for Komodo Tabs
Techniques
ESD Compliance Testing
Electrostatic discharge (ESD) can damage small features and structures in semiconductors and integrated circuits. We offer a comprehensive suite of test equipment which verifies that your devices meet targeted ESD compliance standards.
Learn more ›
ESD Compliance Testing
Electrostatic discharge (ESD) can damage small features and structures in semiconductors and integrated circuits. We offer a comprehensive suite of test equipment which verifies that your devices meet targeted ESD compliance standards.
Learn more ›
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Electron microscopy services for
semiconductors
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Learn more ›
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards