Better understand the properties of your metals and alloys in a faster way
Whether you are developing new alloys or working to improve materials’ properties, you need to be able to increase the accuracy, reproducibility, and speed of the characterization of precipitates and inclusions in metals. Once you do, you can easily achieve trusted results, allowing you to manufacture better materials. However, this involves manual, time-consuming processes that require a high level of image processing expertise.
Thermo Scientific Avizo Software is a versatile analytical solution that provides a high level of accuracy in a fully or semi-automated way for multiple modalities in a single environment. It allows you to dramatically improve your productivity and output on inclusions and precipitates by allowing more people to analyze more samples in a powerful and efficient way—from a single environment.
Avizo Software includes advanced segmentation and quantification tools that allow you to combine various proven segmentation tools, each adding to the previous one for better accuracy. To reduce user bias and ensure your results remain objective, you can design and run repeatable QC workflows to optimize your production processes.
Video Player is loading.
Current Time 0:00
Duration 2:24
Loaded: 0%
Stream Type LIVE
Remaining Time 2:24
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Imaging data
    Workflow using Avizo Software
    Processing
    Workflow using Avizo Software
    Visualization
    Workflow using Avizo Software
    Analysis
    Workflow using Avizo Software
    Customization
    Confidence
    Thanks to powerful segmentation and image processing capabilities and workflows, and after 20+ years of collaboration with the scientific community and thousands of researchers, it has been now proven that our digital imaging-based workflows provide reliable answers to industrial and scientific problems.
    Customization
    Customization
    Because your needs are unique and keep evolving, our software solutions are fully flexible and open to customization. Thanks to our scripting interface (Python, TCL), bridge with MATLAB, and our programming API, you can expand our software solution and integrate your own IP (Intellectual Property). And, if needed, our professional service team can help you design unique solutions tailored to your needs.
    icon-improve_collaboration_64x64
    Support
    Thanks to our dedicated professional support team, you get access to our top experts to ensure that no question is left unanswered. And with our training, consulting options, and ever-expanding collection of tutorials and how-to’s, you can reduce your learning curve and focus on getting the answers you are pursuing.
    Software_Automation_65x64
    Automation
    Thanks to our automation capabilities, and with the addition of our always expanding online repository of add-ons (Xtra Gallery), you can encapsulate repeatable workflows into easy-to-reproduce recipes. With the addition of artificial intelligence, analysis can be performed by non-image processing experts, allowing them to save time on complex analysis while ensuring results consistency. 
    Scientific use cases using Avizo Software
    What is Avizo Software for characterization of metals and alloys?
    During this introductory webinar, you will learn how to:
    • Go from segmenting a dozen particles a day to several hundred 
    • Increase the number of analyzed samples through automation 
    • Remove barriers caused by adopting multiple solutions for day-to-day analysis 
    • Design materials faster and spot weaknesses earlier 
    • Reduce scrap parts and enhance yield; optimize recycling of powder 
    The webinar duration is approx. 20 minutes. The live webinar was followed by a Q&A (question and answer) session. All questions and answers are accessible in the FAQ section.
    Techniques
    Fiber analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of fibrous materials. Avizo Software enables the detection of manufacturing process damage, such as matrix cracking, fiber/matrix debonding, or fiber breakage. Defects in the material can be quantified locally (voids, cracks) or globally (cross-sectional distortions or misalignment).
    Learn more
    Porosity analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of porous materials. Avizo Software enables the detection and classification of various types of porosity (for example, connected vs isolated; macro pores vs micro pores), even on images with complex artifacts (such as pore back effect in FIB-SEM).
    Learn more
    Pore Network Analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of porous materials. This extension enables the conversion of the pore space into a model (Pore Network Model) representing pores and their connectivity, on which various properties can be color mapped. It also provides advanced pore space statistics (pore size distribution, pore throat size distribution, channel length, etc.) and filtering of the model based on its properties.
    Permeability analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of your materials. The Xlab extension provides numerical simulation capabilities to compute absolute permeability, thermal conductivity, electrical resistivity, and molecular diffusivity.
    Failure Analysis
    Avizo Software provides a fast and robust meshing engine to automatically create simulation-ready meshes from 3D labeled images. Avizo Software generates high-quality meshes with smoothed conforming interfaces from large multi-material segmented images. You have fine control over element size distribution, while Avizo Software preserves thin structures and topology. Meshes can be exported to major solver’s format along with boundary conditions. Avizo Software also provides import of the solver’s outputs for post-processing the results with advanced visualization tools.
    Grain Analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of grains. Avizo Software provides advanced tools and enables the detection, segmentation, and analysis of grains, their size, distribution, and shape, even on complex images with artifacts. Grains can then be filtered, classified, and analyzed allowing full size, distribution, and shape statistical extraction. Avizo Software also provides the tools to automate and repeat your analysis through easy-to-customize recipes.
    Particles analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of particles to improve performance of your materials, whether catalysts, alloys, or polymers. Avizo Software provides advanced tools to enable the detection, segmentation, and analysis of particles. Particles can then be filtered, classified, and analyzed, allowing full size, distribution, and shape statistical extraction. Avizo Software also provides the tools to automate and repeat your analysis through easy-to-customize recipes.
    Fiber analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of fibrous materials. Avizo Software enables the detection of manufacturing process damage, such as matrix cracking, fiber/matrix debonding, or fiber breakage. Defects in the material can be quantified locally (voids, cracks) or globally (cross-sectional distortions or misalignment).
    Learn more
    Porosity analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of porous materials. Avizo Software enables the detection and classification of various types of porosity (for example, connected vs isolated; macro pores vs micro pores), even on images with complex artifacts (such as pore back effect in FIB-SEM).
    Learn more
    Pore Network Analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of porous materials. This extension enables the conversion of the pore space into a model (Pore Network Model) representing pores and their connectivity, on which various properties can be color mapped. It also provides advanced pore space statistics (pore size distribution, pore throat size distribution, channel length, etc.) and filtering of the model based on its properties.
    Permeability analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of your materials. The Xlab extension provides numerical simulation capabilities to compute absolute permeability, thermal conductivity, electrical resistivity, and molecular diffusivity.
    Failure Analysis
    Avizo Software provides a fast and robust meshing engine to automatically create simulation-ready meshes from 3D labeled images. Avizo Software generates high-quality meshes with smoothed conforming interfaces from large multi-material segmented images. You have fine control over element size distribution, while Avizo Software preserves thin structures and topology. Meshes can be exported to major solver’s format along with boundary conditions. Avizo Software also provides import of the solver’s outputs for post-processing the results with advanced visualization tools.
    Grain Analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of grains. Avizo Software provides advanced tools and enables the detection, segmentation, and analysis of grains, their size, distribution, and shape, even on complex images with artifacts. Grains can then be filtered, classified, and analyzed allowing full size, distribution, and shape statistical extraction. Avizo Software also provides the tools to automate and repeat your analysis through easy-to-customize recipes.
    Particles analysis
    Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of particles to improve performance of your materials, whether catalysts, alloys, or polymers. Avizo Software provides advanced tools to enable the detection, segmentation, and analysis of particles. Particles can then be filtered, classified, and analyzed, allowing full size, distribution, and shape statistical extraction. Avizo Software also provides the tools to automate and repeat your analysis through easy-to-customize recipes.
    Services
    Amira-Avizo-Introductory-training_1160x600
    Introductory training
    Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
    The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
    Amira-Avizo-advanced-training_1160x600
    Advanced training
    Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
    The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
    Amira-Avizo-Custom-Dev_1160x600
    Custom development
    With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
    We can customize and expand our software solutions at various levels.
    Resources
    Video Player is loading.
    Current Time 0:00
    Duration 1:32
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:32
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • default, selected
      Metallic sample imaged by micro-CT, a digital structure analysis performed with Avizo, to measure influence of volume percentage and particle size on foam's density and cell size. Courtesy of Dr. M. Saadatfar from the Australian National University.
      Video Player is loading.
      Current Time 0:00
      Duration 1:32
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 1:32
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        Metallic sample imaged by micro-CT, a digital structure analysis performed with Avizo, to measure influence of volume percentage and particle size on foam's density and cell size. Courtesy of Dr. M. Saadatfar from the Australian National University.
        Features
        Import and process your imaging data
          • Handle any modality, at any scale, of any size:
          - X-ray tomography: CT, micro-/nanoCT
          - Electron microscopy
          - Synchrotron
          • Support for multi-data/multi-view, multi-channel, time series, very large data
          • Scaling, calibration, conversion, re-sampling
          • Image enhancement, comprehensive filtering and convolution, Fourier frequency transforms
          • Artifact reduction algorithms
          • Advanced multi-mode 2D/3D automatic registration
          • Image stack alignment, arithmetic, correlation, fusion
          Easily segment your imaging data
          • Thresholding and auto-segmentation, object separation, automatic labeling
          • Region growing, snakes, interpolation, wrapping, smoothing
          • Morphological processing, including watershed and basins
          • Machine Learning-based segmentation
          • Automatic tracing of individual fibers and filaments
          • Skeletonization and filament network extraction
          • 3D surface reconstruction
          • Grid generation for FEA/CFD
          Image-to-simulation workflows
          • 3D image-based meshing for Finite Element and CFD
          • Porosity/pore connectivity analysis and skeletonization for Pore Network Modeling
          • Direct 3D image-based simulation: absolute permeability, molecular diffusivity, electrical resistivity, and thermal conductivity computation
          Export your analysis and visualization work to seamlessly publish and present it
          • Animation and video generation
          • Advanced key frame and object animation
          • Mix images, geometric models, measurements and simulations
          • Annotations, measures legends, histograms and curve plots
          • Export spreadsheets, 3D models, high-quality images
          Visualize and explore your imaging data
          • Interactive high-quality volume
          • Orthogonal, oblique, cylindrical and curved slicing
          • Contouring and iso-surface extraction
          • Maximum Intensity or other types of projections
          • Vector and tensor visualization
          Analyze your imaging data and obtain quantitative information
          • Intuitive recipe creation, customization, automated replay
          • Built-in measurements, including counts, volumes, areas, perimeters, aspect ratios and orientations
          • User-defined measures
          • Results viewer with spreadsheet tool and charting
          • Automatic individual feature measurements, 3D localization and spreadsheet selection
          • Automated statistics, distribution graphs
          • Feature filtering using any measurement criterion
          • Data registration, deformation, comparison and measurements
          • Porosity detection and measurement
          • Fiber analysis
          • Pre-processing for structural and flow simulations
          • Import of CAD models for actual/nominal comparison
          Easily and quickly adapt Avizo Software to your specific needs
          • Custom C++ modules development
          • MATLAB™ bridge
          • Python scripting API
          Contact us
          Style Sheet for Global Design System
          Style Sheet for Komodo Tabs
          Style Sheet for Support and Service footer
          Style Sheet for Fonts
          Style Sheet for Cards