Online SPE
Clean-up and concentrate target sample analytes online in minutes
Our online SPE columns contain popular Thermo Scientific HyperSep SPE chemistries to retain specific analytes in a sample matrix. Use these online SPE columns with an appropriate HPLC column and switching valves.
  • Effective removal of contaminants such as proteins from samples
  • Compatible with conventional HPLC systems
  • Fast and effective clean-up and concentration of target compounds
Online Solid Phase Extraction Columns
Thermo Scientific online SPE Columns are available in several convenient formats for sample preparation and provide quick, easy connection by means of a reusable drop-in guard cartridge holder.
  • HyperSep Retain PEP for retention of polar and nonpolar analytes
  • HyperSep Retain CX for retention of basic and nonpolar analytes
  • HyperSep Retain AX for retention of acidic and nonpolar analytes
  • HyperSep Hypercarb for retention of extremely polar analytes
HyperSep Javelin direct-connect online SPE columns
ID (mm)Length (mm)Retain REPRetain CXRetain AXHypercarbQuantity
2.11060310-20160310-30160310-40160310-5014 Pack
3.01060310-20260310-30260310-40260310-5024 Pack
HyperSep UniGuard direct-connect online SPE columns
ID (mm)Length (mm)Retain REPRetain CXRetain AXHypercarbQuantity
2.11060311-20160311-30160311-40160311-5014 Pack
3.01060311-20260311-30260311-40260311-5024 Pack
HyperSep HPLC columns for online SPE
ID (mm)Length (mm)Retain REPRetain CXRetain AXHypercarbQuantity
2.12060312-20160312-30160312-40160312-5011 Each
3.02060312-20260312-30260312-40260312-5021 Each
Ultimate guide to solid phase extraction
This 144-page guide includes:
What you need to know about:Methods for:
  • SPE Phase Selection
  • SPE Workflow
  • Solvent selection
  • Method development & optimization
  • Pharmaceutical/Biotech
  • Environmental
  • Forensic
  • Food Safety
Download guide to solid phase extraction
CMD SchemaApp code