More Filters
System
Description
Dionex IonPac CS21-Fast-4μm Column
Copy link
Application: Determination of cationic polar pesticides using IC-MS/MS
Column features: Fast analysis,High-pressure IC
Eluent: MSA
Dionex IonPac CS20 Column
Copy link
Application: Analysis of inorganic cations and amines
Eluent: MSA
Dionex IonPac CS19-4μm Column
Copy link
Application: Separation of polar amines, moderately hydrophobic amines, polyvalent amines and common cations
Column features: High-pressure IC
Eluent: MSA
Dionex IonPac CS19 Column
Copy link
Application: Separation of polar amines, moderately hydrophobic amines, polyvalent amines and common cations
Eluent: MSA
Dionex IonPac CS18 Column
Copy link
Application: Gradient separation of polyvalent and moderately hydrophobic amines
Eluent: MSA
Dionex IonPac CS17 Column
Copy link
Application: Gradient separation of polyvalent and more hydrophobic amines, biogenic amines and diamines
Eluent: MSA
Dionex IonPac CS16-Fast-4μm Column
Copy link
Application: Analysis of disparate concentration ratios of sodium and ammonium,Analysis of short chain amines and common cations
Column features: Fast analysis,High-pressure IC
Eluent: MSA,Sulfuric acid
Dionex IonPac CS16-4μm Column
Copy link
Application: Analysis of disparate concentration ratios of sodium and ammonium,Analysis of short chain amines
Column features: High-pressure IC
Eluent: MSA,Sulfuric acid
Dionex IonPac CS16 Column
Copy link
Application: Analysis of disparate concentration ratios of adjacent eluting cations,Analysis of short chain amines
Eluent: MSA,Sulfuric acid
Dionex IonPac CS15 Column
Copy link
Application: Determination of disparate concentration ratios of ethanolamine and ammonium
Eluent: MSA,Sulfuric acid
Dionex IonPac CS14 Column
Copy link
Application: Separation of aliphatic amines, alkanolamines, ammonium, alkali, and alkaline earth cations
Eluent: MSA,Sulfuric acid
Dionex IonPac CS12A Column
Copy link
Application: Analysis of inorganic cations and ammonium,Separation of mono- and divalent cations
Column features: Fast analysis
Eluent: MSA,Sulfuric acid
Dionex IonPac CS12A-MS Column
Copy link
Application: Analysis of inorganic cations and ammonium using IC-MS
Column features: Fast analysis
Eluent: MSA,Sulfuric acid
Dionex IonPac CS12 Column
Copy link
Application: Isocratic separation of common inorganic cations
Eluent: MSA,HCl
Dionex IonPac CS11 Column
Copy link
Application: Separation of inorganic cations and aliphatic and aromatic amines
Eluent: HCI/DAP-HCI
Dionex IonPac CS10 Column
Copy link
Application: Separation of inorganic cations and aliphatic and aromatic amines
Eluent: HCI/DAP-HCI
Application: Non-suppressed conductivity detection of inorganic cations, ammonium, select alkanolamines, and transition metals
Eluent: MSA