natural gas detector
Critical information for quick response
Protect workers and the environment from combustibles and toxic gases with equipment that gives you the power to take the appropriate action to minimize hazards. We provide lightweight, intrinsically safe monitoring instrumentation that detects natural gases in diverse applications including US EPA Method 21 monitoring, site remediation, landfill monitoring, and general area surveys.
Featured natural gas detectors
Detect virtually all organic and inorganic compounds. The Thermo Scientific TVA2020 Toxic Vapor Analyzer is a lightweight, intrinsically safe portable detector that can be configured as flame ionization detector (FID) or dual detection technology (FID/PID-photo ionization detector) to offer fast response times.
Make automated measurements of methane and non-methane hydrocarbons with the Thermo Scientific Direct Methane & Non-Methane Hydrocarbon Analyzer, Model 55i. Unlike instruments that measure only methane and total hydrocarbons, this back-flush gas chromatography (GC) system provides a direct measurement of non-methane concentrations.
Support
Let us handle the repair or calibration of all your environmental process and monitoring products. We're happy to track the process from start to finish, including coverage verification, problem diagnosis, cost estimates, and repair costs.
Request service today