Rheology & Extrusion in Food Manufacturing
Food manufacturing quality assurance using rheology.
Food materials in industrial processes often exhibit complex flow properties. Understanding the rheological properties and behavior of food materials under operating conditions is fundamental to ensuring efficient operation and quality food products. This includes food product emulsification, filling, coating, shaping, pumping and bottling processes. Rheometers can determine key properties such as steady shear viscosity, yield stress, viscoelasticity, morphology, droplet size distribution and stringiness.
For quality control, the Thermo Scientific HAAKE Viscotester iQ Rheometer is the ideal partner. Whether it’s a quick single point measurement for batch release or a more comprehensive rheological analysis of the final product, the compact and easy-to-operate Viscotester iQ Rheometer will do the job.
Advance your QA/QC measurements to the next level with Thermo Scientific HAAKE MARS iQ Rotational Rheometer. Measure the viscoelastic properties of foodstuffs like apple sauce and peanut butter to assess texture, mouthfeel, spreadability and more. This modular rheometer offers a range of accessories to help evaluate and verify liquid and semi-solid food formulations throughout the product life-cycle (from production to storage to consumption). 
Read the application note Rheological and Textural Properties of Various Food Formulations Analyzed with a Modular Rheometer Setup.
Thermo Scientific HAAKE MARS Rheometers are designed to measure the broadest range of sample types from low viscous, water-like juices to highly viscoelastic cookie dough, to stiff, paste-like cream cheese or peanut butter. This robust R&D rheometer can obtain all relevant rheological parameters across a wide temperature variance and, if needed, also a wide pressure range.
Video spotlight: Calibrations in HAAKE RheoWin Software
Performing UV curing measurments on the Thermo Scientific HAAKE MARS 40/60 Rheometer
The HAAKE MARS 40 / 60 rheometer is ideally suited for UV curing measurements. Pneumatic release of the upper disposable geometry after measurements allows easy and fast changeover between measurements.
Video Player is loading.
Current Time 0:00
Duration 1:47
Loaded: 9.30%
Stream Type LIVE
Remaining Time 1:47
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
      1. Now Playing
        Up NextPerforming UV curing measurments on the Thermo Scientific HAAKE MARS 40/60 Rheometer
        The HAAKE MARS 40 / 60 rheometer is ideally suited for UV curing measurements. Pneumatic release of the upper disposable geometry after measurements allows easy and fast changeover between measurements.
      2. Now Playing
        Up NextHow to perform a measurement with the RheoRaman System
        How to perform a measurement with the RheoRaman module using the iXR Raman Spectrometer and HAAKE MARS Rheometer.
      3. Now Playing
        Up NextThermo Scientific HAAKE MARS Rheometer Platform
        Material scientists in applied R&D, advanced quality control, and academic research can now use the modular Thermo Scientific HAAKE MARS 40 and 60 rheometers to increase workflow efficiency and minimize operator error while analyzing.
      4. Now Playing
        Up NextThe HAAKE MARS rheometers and HAAKE RheoWin software for Quality Control Labs
        Showing the features that make your rheological measurements in quality control. Precise and reproducible, fast & convenient, safer & more reliable.
      5. Now Playing
        Up NextConducting a Viscosity Calibration in the Thermo Scientific HAAKE RheoWin Software
        Conducting a Viscosity Calibration in the Thermo Scientific HAAKE RheoWin Software
      6. Now Playing
        Up NextCalibrating the Temperature in the Thermo Scientific HAAKE RheoWin Software
        Calibrating the Temperature in the Thermo Scientific HAAKE RheoWin Software
      7. Now Playing
        Up NextConducting a Viscosity Calibration in the Thermo Scientific HAAKE RheoWin Software
        Conducting a Viscosity Calibration in the Thermo Scientific HAAKE RheoWin Software
    Food rheology and extrusion ebooks
    Guide to Skilled Food Rheology & Extrusion
    Understand what to measure in your food development and processing workflows, and which tools are best to measure it.
    Explore two collections of scientific application notes, interactive resources and articles that cover enhancing food properties via rheological measurements and  the development of foods via extrusion.
    Webinar: How extrusion conditions influence the properties of starch compounds
    Watch this webinar to see the benefits of adeptly designing a starch matrix by twin-screw extrusion.
    Learn how to influence the texture, stability and further processability of the final product via extrusion.
    Watch now ›
    HDI-webinar-extrusion-starch-230x195