Add leading x-ray inspection technology to flexible checkweighing solutions to minimize space and integrate operation.  The NextGuard is ideal for finding dense or sharp objects in a wide variety of products as well as detecting errors like over/under fill or missing, misplaced, cracked, or incorrectly formed products. The Versa checkweigher has proprietary software with advanced automatic-zero algorithms to ensure accuracy standards are maintained over time, regardless of environment, and is engineered for ease of maintenance.
x-ray-combo
Features and benefits
Single point of operation for personnel
  • Many companies have quality control procedures where critical control points within their production process need to be monitored. Having the x-ray inspection  combined with the checkweigher, enables both systems to easily be monitored by the employee on the production floor and not have to move a great distance between the two systems. 
  • Reduces safety risk to the operator as the reject systems of both the x-ray system and checkweigher system are located next to each other.
Single point for maintenance
  • Set-up and calibration of the x-ray and checkweigher systems can be done together at the same time.
Space saving
  • Production facilities are often very crowded and constrained by lack of extra space. Even a space savings of 200 cm is valuable within a constricted manufacturing plant. This combination system can save from 200 cm to 1,000 cm in length of valuable packaging line space.
Budget saving
  • Combination units save on the purchase of hardware. Since the conveyor belt of the NextGuard x-ray is also used as the infeed for the Versa checkweigher this eliminates the need for an extra conveyor belt which can be a cost savings of several thousand dollars compared to buying two separate systems.
Single point for quality control
  • Rejection of out-of-range weight and contaminated products are side by side for easy removal. 
  • Each system has its own HMI control screen, and both are visible from a single point allowing simultaneous review of real time data from both systems.
  • X-ray belt can help pull pitch of products, regulating product presentation going onto the checkweigher.
X-ray inspection available as combos
NextGuard C330 X-ray inspection as combo
Sensitivity Very high
Case width
340 mm
Technology
X-ray inspection; product verification; mass estimation
Shielding courtains
Standard or thin-slit
HMI Screens
Both are visible from a single point allowing simultaneous
review of real time data from both systems
Cleaning IP65 with air conditioner; IP54 with fan
Checkweigher combination units
VersaWeigh checkweigher
Versa GP checkweigher
Design
Widest application range
Cantilever design
Weight range
10g to 10kg weighing range, high accuracy
10g to 3kg weighing range, ultra-high accuracy
Throughput*
Up to 400 packs per minute throughput
Up to 400 packs per minute throughput
Material handling
Belt
Belt
Cleaning**
Light wash down, IP65
Light wash down, IP65
* Pack rates dependent upon package length, weigh table length and data output/archiving
** For other cleaning and sanitation requirements contact factory
NextGuard C330 X-ray inspection for combo units
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00