An HID technical applications newsletter
Interact Reference Component
Integrated human identification solutions
When the world seeks the truth, it turns to you. But uncovering the truth presents complex challenges that require a partner you can depend on. That’s why listening to and answering to you is our highest priority, and informs everything we do—so you can focus on making a difference.
Thermo Fisher Scientific offers streamlined, end-to-end workflows for forensic DNA analysis to help you resolve difficult cases and provide investigative leads in a forensically responsible manner. We are committed to delivering unparalleled support and continued innovation to meet your needs today and tomorrow.
Request info
Featured forensic DNA analysis products
Generates lab-quality forensic DNA profiles in only 90 minutes with one minute of hands-on time from single-source samples.
This fully automated robot instrument uses PrepFiler Automated Forensic DNA Extraction Kits to precisely purifying nucleic acids for use in downstream forensic DNA analyses.
The SeqStudio Flex Genetic Analyzer, available in both an 8- and 24-capillary format, incorporates a built-in touchscreen computer, data collection software, and a continuously accessible four-plate deck.
Human identification categories
Rapid DNA analysis
The compact, easy-to-use RapidHIT ID System generates forensic DNA profiles in virtually any setting in as little as 90 minutes—and no technical expertise is required. Safer communities are within reach.
Next-generation sequencing
The Precision ID NGS System for HID can help you solve tough cases by getting more information from your challenging samples. Now you have help moving those unknown samples from storage to investigative leads.
Sample preparation
Forensic DNA samples are among the most difficult specimens to process because they are often limited in quantity, may be environmentally exposed, and may be on substrates that contain PCR inhibitors. The quantity and quality of genomic DNA extracted and purified from a forensic sample is directly correlated to the success of downstream analysis.
Automated sample preparation
Easily process up to 96 DNA samples in less than 90 minutes to get more critical answers faster. Reduce case backlogs that delay answers and put lives on hold. The high-throughput HID NIMBUS Presto System can process most complex forensic sample types—including bone, tooth, and touch DNA samples—quickly and easily. Reclaim your time and keep answers flowing with an automated, set-up-and-go system.
DNA quantification
The quantification step of the casework workflow is the key to determining how a sample will be processed downstream. There are many factors that determine the success of the short tandem repeat (STR) profile. Our Quantifiler DNA quantification kits enable forensics labs to achieve accurate results in less time for even the most challenging casework samples.
PCR amplification
Applied Biosystems STR amplification kits combine the sensitivity of PCR with the power of multicolor fluorescence technology to enable robust, reliable results. These forensic DNA STR amplification kits provide maximum commonality in markers to make more cross-border identifications.
DNA analysis (instrument systems)
Applied Biosystems capillary electrophoresis (CE) instrument systems have been validated by the human identification community and are the most widely trusted CE systems available.
Data interpretation (software)
GeneMapper ID-X Software v1.7, an automated genotyping software solution, streamlines human identification data analysis needs, including forensic casework, databasing, and paternity testing. It is an Expert System approved for use on offender samples and known reference samples by National DNA Index System (NDIS).
Challenging sample analysis
Crime scene samples can originate from harsh environmental locations, which may result in various stages of degradation and contamination. Imagine a 20-year-old stamp that was stored away in a closet, trace amounts of blood from an exploded IED, or a partially eaten piece of pizza. How would you begin to evaluate these challenging samples?.
Sexual assault casework
Sexual assault kit (SAK) samples are among the most difficult sample types encountered by many forensic laboratories. Samples collected from female victims may contain very low amounts of male DNA mixed with high amounts of female DNA. It is critical to be able to quickly and accurately assess whether swab evidence from an SAK contains a male contributor before labor-intensive differential extraction procedures are performed.
Validation and verification services
Leave every detail of validating and implementing the forensic DNA testing solutions your work demands to us. Our experienced HID Professional Services (HPS) team can manage any or all parts of the process—including training—to get your new workflow operational quickly.
Training
Training is the key to success. Forensic DNA testing constantly evolves with new technologies, procedures, and requirements. To maintain success, DNA analysts must learn how to best adapt laboratory processes and improve their technical understanding of analysis procedures.
From our trusted PCR amplification kits, genetic analyzers, powerful NGS solutions, and software to in-depth training, on-site technical assistance, and validation services, our integrated systems work to help ensure your success and help maximize your productivity.
Sample collection
Thermo Fisher Scientific is proud to provide innovative solutions, including NUCLEIC-CARDs, Copan 4N6FLOQSwabs, and more, through a collaboration with Copan Italia s.p.a. In combination with our integrated human identification analysis workflows, these collection devices enable forensic and paternity laboratories to maximize efficiency and improve downstream STR analysis and first pass success rate.
Robust PrepFiler extraction kits enable inhibitor removal and help improve the overall yield, concentration, and purity of DNA isolated from both routine and challenging samples. This provides greater quantities of highly purified DNA for improved downstream success.
Learn more ›
Advanced quantification kits that enable forensic labs to achieve accurate results in less time, for even the most challenging casework samples. The kits evaluate sample quality and quantity in a single reaction.
Learn more ›
The QuantStudio 5 Real-Time-PCR System for HID is an easy-to-use, reliable, and affordable real-time PCR system. When used with our latest advances in quantification chemistry and Applied Biosystems HID RealTime PCR Analysis Software, this system offers a sensitive, robust solution for forensic DNA quantification.
Learn more ›
Kits that combine the sensitivity of PCR chemistry with the power of multicolor fluorescence technology to enable robust, reliable results for all human identification applications—even with mixed, degraded, and challenged samples.
Learn more ›
Whether you're processing routine samples, highly degraded samples, or urgent request cases, Thermo Fisher Scientific provides best-in-class DNA analysis solutions to the forensic community. Choose from the proven performance of our family of genetic analyzers, the power of NGS, or the simplicity of rapid DNA to get more answers from your samples as efficiently as possible.
Learn more ›
Specialized software solutions tailored for forensic DNA labs, designed to optimize workflows using advanced algorithms developed and refined over decades of DNA chemistry and instrumentation expertise, enhancing operational efficiency and ensuring the reliability and precision of both raw and processed data.
Learn more ›
Sample collection
Thermo Fisher Scientific is proud to provide innovative solutions, including NUCLEIC-CARDs, Copan 4N6FLOQSwabs, and more, through a collaboration with Copan Italia s.p.a. In combination with our integrated human identification analysis workflows, these collection devices enable forensic and paternity laboratories to maximize efficiency and improve downstream STR analysis and first pass success rate.
Robust PrepFiler extraction kits enable inhibitor removal and help improve the overall yield, concentration, and purity of DNA isolated from both routine and challenging samples. This provides greater quantities of highly purified DNA for improved downstream success.
Learn more ›
Advanced quantification kits that enable forensic labs to achieve accurate results in less time, for even the most challenging casework samples. The kits evaluate sample quality and quantity in a single reaction.
Learn more ›
The QuantStudio 5 Real-Time-PCR System for HID is an easy-to-use, reliable, and affordable real-time PCR system. When used with our latest advances in quantification chemistry and Applied Biosystems HID RealTime PCR Analysis Software, this system offers a sensitive, robust solution for forensic DNA quantification.
Learn more ›
Kits that combine the sensitivity of PCR chemistry with the power of multicolor fluorescence technology to enable robust, reliable results for all human identification applications—even with mixed, degraded, and challenged samples.
Learn more ›
Whether you're processing routine samples, highly degraded samples, or urgent request cases, Thermo Fisher Scientific provides best-in-class DNA analysis solutions to the forensic community. Choose from the proven performance of our family of genetic analyzers, the power of NGS, or the simplicity of rapid DNA to get more answers from your samples as efficiently as possible.
Learn more ›
Specialized software solutions tailored for forensic DNA labs, designed to optimize workflows using advanced algorithms developed and refined over decades of DNA chemistry and instrumentation expertise, enhancing operational efficiency and ensuring the reliability and precision of both raw and processed data.
Learn more ›
Our longstanding commitment to the highest manufacturing standards help ensure you get answers you trust. Read about our quality manufacturing, including our ISO 18385–ready facilities.
Customer videos
Video Player is loading.
Current Time 0:00
Duration 6:58
Loaded: 2.36%
Stream Type LIVE
Remaining Time 6:58
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Combating human trafficking with DNA analysis
    Dr. Bruce Budowle, the  Director of  The Center for Human Identification at the  University of North Texas describes how the Center is developing programs and utilizing DNA technologies to combat human trafficking.
    Video Player is loading.
    Current Time 0:00
    Duration 7:31
    Loaded: 2.18%
    Stream Type LIVE
    Remaining Time 7:31
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Harnessing the power of forensic DNA analysis to solve crimes faster
      Dave Reichert, former King County Sherriff, discusses the decades- long investigation into the Green River Killer and how DNA analysis may help solve crimes faster in the future.
      Video Player is loading.
      Current Time 0:00
      Duration 5:59
      Loaded: 2.74%
      Stream Type LIVE
      Remaining Time 5:59
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Developing a new national database: Validation, standardization and quality requirements
        Police Lieutenant Colonel Lorna Santos describes how Philippines is instituting their forensic DNA database and implementing quality standards to aid in solving crime.
        Video Player is loading.
        Current Time 0:00
        Duration 10:51
        Loaded: 1.52%
        Stream Type LIVE
        Remaining Time 10:51
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Rapid DNA: Forging Partnerships
          Mark Smith, Supervising Forensic Scientist and Rapid DNA Coordinator, describes how AZ Dept of Public Safety took their vision to forge lasting partnerships between the crime lab and their various law enforcement agencies and turned it into reality using Rapid DNA technology.
          Visistat reference component