quantstudio-5-real-time-pcr-system-human-identification-showcase-image-230x195
QuantStudio 5 Real-Time PCR System for Human Identification
Reliable answers, significant savings
The modern forensics laboratory is undergoing a technological revolution. This revolution is driving lab instruments to become increasingly more efficient, reliable, interactive, future-proof, and economical.
As your trusted partner, we understand the need for a reliable performer that delivers. The Applied Biosystems QuantStudio 5 Real-TmePCR System for Human ldentification is a robust and reliable real-time PCR instrument.
This instrument is designed for both new and experienced users who need a simple, reliable, and affordable real-time PCR system that doesn’t require compromise on performance or quality. Used with our latest advances in quantification chemistry and HID real-time PCR software, this system offers a sensitive, robust solution for forensic DNA quantification. It also helps prepare laboratories for future quantification technologies.
With maximum dye versatility, this HID solution offers accurate, trusted results in a small benchtop footprint.
Get a quote
Increased sample assessment for optimal downstream results
The quantification step of the casework workflow is the key to determining how a sample will be processed downstream. There are many factors that determine the success of the short tandem repeat (STR) profile. Our advanced quantification kits enable forensics labs to achieve accurate results in less time for even the most challenging casework samples.
The Applied Biosystems Quantifiler HP and Quantifiler Trio DNA quantification kits are the first kits that enable forensics laboratories to simultaneously obtain a quantitative and qualitative assessment of total human DNA in a single, highly sensitive real-time PCR reaction.
The value of DNA quantification includes:
  • Improved amplification efficiency
  • Fewer off-scale, over-amplified, or under-amplified samples
  • Normalized profiles result in reliable data interpretation
  • Reduced need for downstream reanalysis helps provide long-term cost savings
  • Provides key sample information to enable forensic workflow decisions
  • Enables conservation of sample through targeted downstream assay decisions
Integrated forensic workflow
Integrated-forensic-workflow-740
Featured video
Video Player is loading.
Current Time 0:00
Duration 6:54
Loaded: 0%
Stream Type LIVE
Remaining Time 6:54
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Building a forensic DNA analysis lab
    Liz Peters from Fiji Police Force discusses how bringing new forensic DNA technology into her lab has improved sample processing to help solve crime faster.
    Solutions for DNA quantification
    quantification-kits-showcase-image-230x195
    The Quantifiler Trio and HP kits, used in conjunction with the QuantStudio 5 system and Applied Biosystems HID Real-Time PCR Analysis Software, help redefine the casework sample-processing workflow. Quality functionality enables a more comprehensive view of sample quality (both degradation and inhibition), while increased sensitivity and robustness correlate with more recently released Applied Biosystems STR chemistries. This enables you to screen samples and make critical workflow decisions (for example, autosomal vs. Y-STR kits for capillary electrophoresis (CE), or CE vs. NGS technology) with greater confidence.
    The quantity and quality indicators that these kits incorporate help deliver accurate, sensitive, and specific results that work together with the software to enable informative, integrated workflow decisions.
    The QuantStudio 5 system is designed for both new and experienced users who need a simple, reliable and affordable real-time PCR system without compromising performance or quality.
    agrigenomics-qpcr-quantstudio-image-230x195-Recovered
    quantstudio-5-real-time-pcr-system-human-identification-software-showcase-image-230x195
    We’ve designed software specifically for the HID application to help forensics scientists take full advantage of the data and capabilities offered by the Quantifiler kits, and to help maximize sample-processing efficiency.
    Applied Biosystems HID Real-Time PCR Analysis Software with new virtual standard curve functionality provides an integrated solution for DNA quantification. This software also introduces the capability to run Quantifiler kits on the QuantStudio 5 System. 
    For Research Use Only. Not for use in diagnostic procedures.