• Viabilities routinely >80%
  • Characterized for phase I and phase II enzyme activities
Our rat hepatocytes are prepared using the same careful isolation and cryopreservation techniques as our human lots.  Characterization methods of cryopreserved hepatocytes include ECOD, 7-HCG and 7-HCS for phase I and phase II enzyme activities.  We also closely monitor cell morphology, attachment efficiency, monolayer confluency (plateable cells) and viability stability over time for cryopreserved hepatocytes.
Click below to see our Inventory Data Table to select lots that meet your specific needs
Cryopreserved Hepatocytes Ordering Information
For Research Use Only. Not for use in diagnostic procedures.