Graphic of a bioreactor and a chromatogram
What is process scale chromatography?
Process scale chromatography is a scalable cGMP manufacturing technique for the separation of biomolecules from a feedstock or slurry to produce biotherapeutics. This purification technique can be used in a variety of applications from gene therapies to biosimilars and bio-betters to vaccines and more. It is a cost effective and efficient purification solution that enables high purity and high yield of the target molecule under optimal conditions.
 Contact us
When you need high performance and high throughput on a large scale, POROS bulk chromatography resins enable outstanding performance for process-scale bioseparations. Utilized in multiple processes, these breakthrough resins combine optimal particle size and pore structure with a rigid backbone and proprietary chemistries. POROS 50 µm resins demonstrate exceptionally high resolution, dynamic binding capacity, and capture efficiency over a range of flow rates and are primarily used in polish applications.
CaptureSelect affinity resins offer excellent selectivity resulting in higher yields and improved purity in the initial capture step. We have developed numerous products for large-scale purification of various biologics such as antibodies and antibody fragments, fusion proteins, hormones, and viruses.
Learn more about how to pack POROS chromatography resins ›
Non-affinity resins
Ion exchange chromatography resin and hydrophobic interaction chromatography resin
Custom resins
Custom chromatography services for unique molecules
Packing rigid POROS chromatography resins:
efficient and fast method to get optimal results.
Watch how to pack POROS chromatography resins.
Video Player is loading.
Current Time 0:00
Duration 21:09
Loaded: 0%
Stream Type LIVE
Remaining Time 21:09
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Chromatography Learning Lab
    Check out the new Chromatography Learning Lab from Thermo Fisher Scientific, providing a robust range of webinars, articles, eBooks, infographics, and other digital resources to help you optimize the use of our purification products in your process.
    Explore digital resource
    Bioprocessing resources
    Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
    Pharmaceutical Grade Reagent. For Manufacturing and Laboratory Use Only.