Resin manufacturer
Custom chromatography resin development
Through its unique product attributes, POROS chromatography resins help maintain performance and add flexibility to your downstream process. Combine high resolution and high capacity in a custom chromatography resin tailored to your specific process requirements.
Contact us  Technical resources
Resin development includes the use of Design of Experiments (DoE) to define the resin-manufacturing process required for your purification needs. DoE screening includes pore size, ligand density, and coupling chemistry to optimize performance for your biotherapeutic. Start the process with your ligand or another commercially available ligand, select from our library of existing CaptureSelect ligands, or generate a custom CaptureSelect ligand for your application.
All process validation and regulatory requirements are fully supported by an ISO 9001/13845 quality management system.
Custom POROS resin development program
Prototype resin development
Resin beads for prototype resin development
  • Customer information
  • Experimental design
  • Proposal
Process definition
Prepacked chromatography columns
  • Process optimization
  • Scale-up
Commercialization
bioprocess analytics
  • Process validation
  • Stability studies
  • Regulatory support package
 Arrow
Request a technical meeting
meeting icon
Unlock the power of POROS chromatography, and develop a platform purification process for any biotherapeutic. To arrange a technical meeting with our resin development scientists; simply submit your request for more information and our local sales representative will follow-up with you to organize the meeting.
Chromatography Learning Lab
Check out the new Chromatography Learning Lab from Thermo Fisher Scientific, providing a robust range of webinars, articles, eBooks, infographics, and other digital resources to help you optimize the use of our purification products in your process.
Explore digital resource
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: