Cell Factory systems
Maximize productivity with an integrated solution for Cell Factory systems
Take your Nunc Cell Factory systems to the next level. Nunc Cell Factory equipment delivers the solution for quick and easy scale-up of your existing Cell Factory systems. Nunc Cell Factory equipment automates the filling, emptying, harvesting, and trypsinization steps of adherent cell culture. Realize process consistency, optimization, and standardization when scaling up and manipulating multiple Cell Factory systems simultaneously with this trusted equipment. All equipment is compliant with cGMP.
Request information   Download product brochure
Featured Cell Factory automation equipment products
The Thermo Scientific Nunc Cell Factory manipulator is a fully programmable manipulator that automates the handling of the Nunc Cell Factory systems. Simplify and achieve constant and reliable processes.
The Thermo Scientific Nunc Cell Factory shaker system offers an automated solution for the detachment of cells from Nunc Cell Factory systems; it eliminates the need for shaking and pounding, and supports workflow efficiency.
The Thermo Scientific Nunc Cell Factory incubator is a mobile solution that accommodates four racks of Cell Factory 40-layer or 10-layer systems. it is available with and without CO2 control.
The Thermo Scientific Nunc Cell Factory Racks and Carts integrate perfectly with all Nunc Cell Factory equipment. Available for use with Cell factory 40-, 12-, and 10-layer systems.
Cell Factory automation equipment
Thermo Scientific Nunc Automatic Cell Factory Manipulator System (ACFM)
Increase processing speed by eliminating manual filling, emptying, harvesting and trypsinization steps of all Cell Factory systems. Designed for large-scale cell culture and production of vaccines, cell therapeutics or other biological products.
Video Player is loading.
Current Time 0:00
Duration 4:29
Loaded: 0%
Stream Type LIVE
Remaining Time 4:29
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextThermo Scientific Nunc Automatic Cell Factory Manipulator System (ACFM)
        Increase processing speed by eliminating manual filling, emptying, harvesting and trypsinization steps of all Cell Factory systems. Designed for large-scale cell culture and production of vaccines, cell therapeutics or other biological products.
      2. Now Playing
        Up NextThermo Scientific Nunc Cell Factory Incubator
        Mobile solution for the incubation of Nunc Cell Factory systems, available with and without CO2 control.
      3. Now Playing
        Up NextThermo Scientific Nunc Cell Factory Shaker
        Automate the detachment of cells from Nunc Cell Factory Systems by using the Nunc Cell Factory Shaker System.
    Related products
    Compact, multi-layer single-use cell culture systems designed to meet the needs of scale-up and production of your products.
    Bring your bioproduction processes together seamlessly with an extensive range of standard and customized fluid transfer products.
    Cell Factory ordering information
    Bioprocessing resources
    Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: