Search Thermo Fisher Scientific
Configurations offering both spectral analysis and conventional compensation range from 5 to 9 lasers and up to 60 parameters, while configurations offering only traditional compensation come with 4 or 5 lasers and up to 35 parameters. Other than lasers and detectors, all configurations share the same specifications.
All configurations of the Bigfoot Cell Sorter offer two or more forward and side scatter channels in addition to the fluorescence channels shown in the tables.
Lasers |
Number of fluorescence detection channels for included lasers |
Scatter detectors |
Total detection channels |
Cat. No. |
||||||||
UV |
Violet |
Blue violet |
Blue |
Green |
Yellow green |
Red |
Far red |
Near- |
||||
5 |
12 |
12 |
|
7 |
|
12 |
|
5 |
|
5 |
53 |
|
6 |
12 |
12 |
|
7 |
|
12 |
|
5 |
3 |
5 |
56 |
|
6 |
12 |
12 |
4 |
7 |
|
12 |
|
5 |
|
5 |
57 |
|
7 |
12 |
12 |
4 |
7 |
|
12 |
|
5 |
3 |
5 |
60 |
|
9 |
12 |
12 |
4 |
7 |
12 |
5 |
3 |
5 |
60 |
*532 nm and 594 nm lasers are not used for spectral analysis, but all fluorescence detectors are used.
Lasers |
Number of fluorescence detection channels for included lasers |
Scatter detectors |
Total detection channels |
Cat. No. |
||||||||
UV |
Violet |
Blue violet |
Blue |
Green |
Yellow green |
Red |
Far red |
Near- |
||||
4 |
7 |
|
7 |
|
7 |
|
4 |
|
2 |
27 |
||
5 |
7 |
7 |
|
5 |
|
7 |
|
4 |
5 |
35 |
||
4 |
7 |
|
7 |
|
7 |
|
4 |
|
3 |
28 |
Excitation lasers | 349 nm, 405 nm, 445 nm, 488 nm, 532 nm, 561 nm, 594 nm, 640 nm, 785 nm |
Optical power | Free space delivery of 349 nm (100 mW), 405 nm (100 mW), 445nm (200 mW), 488 nm (125 mW), 532 nm (100 mW), 561 nm (100 mW), 594 nm (100 mW), 640 nm (100 mW), 785 nm (100 mW) |
Beam alignment | Fixed, 7 spatially separated pinholes |
Detection parameters | up to 55 fluorescence + 5 scatter |
Scatter parameters | Standard FSC and SSC 488 nm; small particle FSC 405 nm; depolarized FSC and SSC 488nm |
Scatter resolution | <0.2 µm scatter resolution from background with small particle detector |
Pulse measurement | Simultaneous measured peak, area, and width for sample input and output of every channel |
Fluorescence sensitivity | <100 MESF for FITC, PE, APC |
Nozzles | 70, 100, 120, and 150 µm ceramic nozzle tips with adjustable pressure settings |
Sample input tube types | 1.5 mL, 5 mL, and 15 mL |
Temperature control | 4°C-37°C for input and output of all media types |
Agitation | Automatic based on input tube type |
Event rate | >100,000 eps acquisition with all 60 parameters |
Sorting speed | >70,000 eps with all parameters enabled* |
Sort devices | Up to 6-way sorting; configurable tube holders include 1.5 mL, 5 mL, 15 mL, and 50 mL tubes. Microwell plates with a standard footprint of up to 1,536 wells can be loaded directly for straight-down sorting; 4-way sorting into 96-well plates; 4-way sorting into 384 well plates. Temperature control from 4°C to 37°C for all sorting media. |
* When running at 70,000 eps, users may see a decrease in efficiency depending on experimental conditions and sort criteria. Sort speeds should always be optimized for specific experiments.
Compliance testing | The Bigfoot Spectral Cell Sorter biosafety enclosure provides personnel and product protection similar to a Class II BSC. Test procedures and criteria laid out within NSF49 and EN12469 standards can be utilized to demonstrate performance. |
Dimensions | 99 cm W x 99 cm D x 170 cm H (39 in W x 39 in D x 67 in H) |
Fluidics cart | None |
Weight | 1433 lbs (650kg) |
Monitor | 32-inch, 4k |
Power supply | 100-240VAC 50/60Hz |
Power consumption | 750W max (instrument, PC and monitor) |
Relative humidity | 20-60% RH |
For Research Use Only. Not for use in diagnostic procedures.