There are many options available for protein expression from cloned DNA. These include cell-free extracts (in vitro expression systems), and bacterial, yeast, insect, and mammalian cell systems, each with its own advantages and drawbacks. Requirements for protein solubility, functionality, and yield are often the most important factors to consider when choosing an expression system, but the time and labor required for subcloning and optimizing expression can also have a significant impact.
Use of recombinant proteins varies widely - from functional studies in vivo to large-scale protein production for structural studies and therapeutics. Using the best expression system for your protein and application is key to your success.
Read about protein expression optimization
Get protein expression basics support 
The following table highlights the characteristics of the commonly-used expression hosts.
Select the best expression host for your research
Host organismTypical applicationsAdvantagesPotential Challenges
Cell-free
  • Rapid expression screening
  • Expression of toxic proteins
  • Incorporation of unnatural labels or amino acids
  • Functional assays
  • Protein interaction studies
  • Rapid expression directly from plasmid
  • Open system—easily add components to enhance protein solubility or functionality
  • Simple format
  • Scalable
  • Compatible with Gateway cloning
  • Difficult to obtain protein yields >3 mg
Bacteria
  • Structural analysis
  • Antibody generation
  • Functional assays
  • Protein interaction studies
  • Scalable
  • Low cost
  • Simple culture conditions
  • Compatible with Gateway cloning
  • Protein solubility
  • Minimal post-translational modifications possible
  • May be difficult to express functional mammalian proteins
Yeast
  • Structural analysis
  • Antibody generation
  • Functional assays
  • Protein interaction studies
  • Eukaryotic protein processing
  • Scalable, with very high yields possible using fermentation (g/L)
  • Simple media requirements
  • Fermentation required for very high yields
  • Growth conditions may require optimization
Insect
  • Functional assays
  • Structural analysis
  • Antibody generation
  • Post-translational modifications similar to mammalian systems
  • Higher yield than mammalian systems
  • Compatible with Gateway cloning
  • Relatively demanding culture conditions
Mammalian
  • Functional assays
  • Protein interaction studies
  • Antibody generation
  • Highest level of correct post-translational modifications possible
  • Highest probability of obtaining fully functional human proteins
  • Compatible with Gateway cloning
  • Multimilligram per liter yields only possible in suspension cultures
  • Relatively demanding culture conditions
Protein Expression with Gateway technology
Gateway technology allows highly efficient transfer of the gene of interest into any of a number of expression vectors from a single entry clone. The variety of expression choices available makes the Gateway system of recombination cloning ideal for protein expression studies.  
The Gateway recombination cloning system is designed for highly efficient transfer of your DNA into any Gateway expression vector from a single entry clone. The extremely wide variety of Gateway cloning–compatible expression vectors available makes the Gateway system of recombination cloning ideal for protein expression studies.
  • One hour, room temperature cloning reactions with up to >99% efficiency deliver the clone you need
  • Maintain orientation and reading frame without using restriction enzymes or ligation to make expression-ready clones
  • Use the same clone from target identification to validation to help ensure consistent results throughout your experiment, without re-sequencing
  • Easily shuttle insert DNA from one expression vector to another—get flexibility while simplifying your cloning workflow
  • Learn more about Gateway cloning
For T7-regulated E. coli expression systems, MagicMedia Expression Medium can increase protein yields up to 10-fold. The exclusive MagicMedia formulation enables E. coli to reach culture densities 3–10-fold higher than those achieved with traditional LB + IPTG media.
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.