TOPO XL-2 Complete PCR Cloning Kit
TOPO XL-2 Complete PCR Cloning Kit
Confidently conquer cloning of long DNA fragments
Long-fragment cloning can be challenging for many reasons including the limitations of traditional molecular biology reagents. The Invitrogen TOPO XL-2 Complete PCR Cloning Kit provides all the necessary elements to overcome these limitations and enable precise, efficient, and simple cloning of long PCR products up to 13 kb. You can achieve higher cloning efficiencies in just 5 minutes at your benchtop when utilizing the new linearized and topoisomerase I–activated pCR-XL-2-TOPO vector, which is compatible with high-fidelity blunt-end PCR fragments produced by the Invitrogen Platinum SuperFi Green PCR Master Mix.
Download flyer
Kit contents
The Complete PCR Cloning Kit includes:
  • TOPO XL-2 PCR Cloning Kit—contains the pCR-XL-2-TOPO vector
  • Platinum SuperFi Green PCR Master Mix—features a proofreading DNA polymerase (with >300x the fidelity of Taq polymerase) with high processivity to generate accurate, long amplicons, and an inert density ingredient that allows you to load directly onto an agarose gel for gel resolution and extraction
  • PureLink Quick Gel Extraction and PCR Purification Combo Kit—designed to purify DNA fragments in less than 30 minutes from agarose gels or direct PCR purification using a silica-based spin cartridge
  • One Shot OmniMAX 2 T1R Chemically Competent E. coli Cells—an improved high-efficiency chemically competent cell line, perfect for use in all cloning applications
The pCR-XL-2-TOPO vector
Figure 1. The pCR-XL-2-TOPO vector.
The pCR-XL-2-TOPO vector (Figure 1) includes:
  1. ccdB gene for positive selection
  2. EcoRI site flanking the PCR product insertion site for easy excision of inserts
  3. Ampicillin and kanamycin resistance genes for your choice of antibiotic selection
  4. T7 promoter/priming site for in vitro transcription
  5. T7, T3, and M13 forward and reverse primer sites for sequencing
Ordering information
Advantages of the TOPO XL-2 Complete PCR Cloning Kit
The TOPO XL-2 Complete PCR Cloning Kit is optimized to provide the highest cloning efficiencies for extra-long PCR fragments, as illustrated in Figures 2, 3, and 4.   
The TOPO XL-2 Complete PCR Cloning Kit has superior cloning efficiency for a broad range of different sizes of targets.
Figure 2. TheTOPO XL-2 Complete PCR Cloning Kit has superior cloning efficiency for a broad range of different sizes of targets. Lambda genomic DNA PCR fragments ranging from 1 kb to 13 kb were cloned using the TOPO XL-2 cloning kit.
The TOPO XL-2 cloning kit shows superior cloning efficiency compared to the TOPO XL kit.
Figure 3. The TOPO XL-2 cloning kit shows superior cloning efficiency compared to the TOPO XL kit. Recommended protocols for each kit were followed for cloning a 7 kb control insert.
The TOPO XL-2 cloning kit has superior cloning efficiency compared to an In-Fusion® Cloning kit (Clontech) over a broad range of target sizes.
Figure 4. TheTOPO XL-2 cloning kit has superior cloning efficiency compared to an In-Fusion® Cloning kit (Clontech) over a broad range of target sizes. Human and lambda genomic DNA targets were cloned following the protocols given in the respective kits’ manuals. Cloning efficiency was determined by colony PCR using an internal primer that spans either a 3 kb (human) or ~5 kb (lambda) region of the cloned targets. The high-fidelity polymerase enzyme from the In-Fusion Cloning kit was unable to amplify the human 10 kb DNA target. (See Figure 7 in “Improved sequence accuracy and specificity of inserts”.)
Platinum SuperFi DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with the trusted Invitrogen Platinum hot-start technology, designed for the highest success in PCR. Featuring >300x the fidelity of Taq polymerase, Platinum SuperFi Green PCR Master Mix, included in the TOPO XL-2 PCR Cloning Kit, is ideally suited for cloning, mutagenesis, and other applications benefiting from superior sequence accuracy, such as working with long DNA fragments.
Platinum SuperFi DNA Polymerase exhibits superior fidelity
Figure 5.Platinum SuperFi DNA Polymerase exhibits superior fidelity. Polymerase fidelity was measured by next-generation sequencing using unique molecular identifiers (UMI), and reads from the same UMI family were aligned to call errors. The polymerase fidelities were normalized to that of Taq polymerase.
Platinum SuperFi DNA Polymerase has higher specificity in amplifying larger targets than the Invitrogen Elongase Enzyme Mix.
Figure 6.Platinum SuperFi DNA Polymerase has higher specificity in amplifying larger targets than the Invitrogen Elongase Enzyme Mix. Targets of 1 kb, 3 kb, 7 kb, 10 kb, and 13 kb in length from lambda genomic DNA were amplified following the manufacturers’ standards for PCR cycling parameters. The Elongase Enzyme Mix is recommended for the TOPO XL PCR Cloning Kit, and Platinum SuperFi DNA Polymerase is provided with the TOPO XL-2 Complete PCR Cloning Kit.
Platinum SuperFi DNA Polymerase shows greater specificity for both human and lambda gDNA targets than Clontech’s high-fidelity (HiFi) polymerase.
Figure 7.Platinum SuperFi DNA Polymerase shows greater specificity for both human and lambda gDNA targets than Clontech’s high-fidelity (HiFi) polymerase. Amplification protocols were followed per manufacturers’ recommendations. From a 50 µL PCR reaction, 6.5 µL of the human gDNA targets and 4 µL of the lambda gDNA targets were loaded into a 1% agarose gel stained with EtBr.
The PureLink Quick Gel Extraction and PCR Purification Combo Kit offers the ability to quickly perform both a gel extraction and a PCR purification with a single kit. The isolated DNA is then ready to use for cloning. By including the PureLink kit in the TOPO XL-2 Complete PCR Cloning Kit, the total yield of DNA is greatly improved due to less handling, a major advantage when working with larger DNA fragments (Figure 8).
The PureLink Quick Gel Extraction and PCR Purification Combo Kit shows greater total DNA yields than the Invitrogen S.N.A.P. Gel Purification Kit, even for larger fragment sizes.
Figure 8. The PureLink Quick Gel Extraction and PCR Purification Combo Kit shows greater total DNA yields than the Invitrogen S.N.A.P. Gel Purification Kit, even for larger fragment sizes. Gel-purified lambda PCR products of various sizes were amplified with Platinum SuperFi 2X Green Master Mix. The same starting material was used for equivalent gel purification using PureLink (provided with TOPO XL-2 kits) and S.N.A.P. (provided with TOPO XL kits) kits, for comparison. Purification protocols were followed as stated in the manuals. Total DNA yield amounts were quantified and compared. Percentages indicate the increase in yield obtained from the PureLink kit compared to the S.N.A.P. kit.
OmniMAX 2 T1R competent cells offer the highest transformation efficiency (> 5 x 109 transformants/µg pUC19) of any chemically competent E. coli cells in the Invitrogen One Shot format (Figure 9). These highly versatile cells also provide efficient transformation of highly methylated DNA, since OmniMAX 2 T1R cells lack the E. coli K12 restriction systems (mcrA Δ(mrr hsdRMS mcrBC)). In addition, the strain has the tonA genotype, which confers resistance to T1 and T5 phage infection. This protects your samples and minimizes the possibility of downtime in your lab due to phage contamination.
The chemically competent OmniMAX™ 2 T1R cells yield higher numbers of colony forming units (cfu) than TOP10 cells, following transformation of cloned DNA fragments.
Figure 9.The chemically competentOmniMAX 2 T1R cells yield higher numbers of colony forming units (cfu) than TOP10 cells, following transformation of cloned DNA fragments. Clones were plated on selection plates with Kanamycin for both OmniMax 2 T1R and TOP10 cells, and with 1 mM IPTG for just the OmniMAX 2 T1R cells. Ten to twelve transformants were analyzed by colony PCR.
Helpful Hint
Plate OmniMAX 2 T1R cells with 1 mM IPTG added to the plating medium to reduce background from empty vector, therefore facilitating screening and selection.
Invitrogen SYBR Safe DNA Gel Stain is the recommended gel stain. It offers the highest sensitivity by utilizing blue light, eliminating the damaging effects of UV light exposure (Figure 10). While ethidium bromide (EtBr) is still a sensitive stain, the use of UV light can nick DNA and reduce cloning efficiency. Crystal violet stain utilizes white light and therefore does not damage the DNA; however, it is less sensitive.
Ethidium bromide (EtBr) and SYBR Safe DNA Gel Stain are more sensitive than crystal violet.
Figure 10.Ethidium bromide (EtBr) and SYBR Safe DNA Gel Stain are more sensitive than crystal violet.  HML = High DNA Mass Ladder.
Support
    TOPO Cloning Technology
    Learn more about our TOPO Cloning technology and other TOPO Cloning kits we offer
    Cloning Support Center
    Find tips, troubleshooting help, and resources for your cloning applications
    Cloning Education
    Learn about PCR cloning strategies and subcloning approaches
    PCR Education
    Review the considerations for long-fragment PCR, reaction setup, and more
    For Research Use Only. Not for use in diagnostic procedures.