Lab Automation Incubators
Robot-friendly storage and incubation at 37°C
Achieve extremely uniform environmental conditions for all microplates, elimination of recovery times, and reduced assay variance. The Thermo Scientific Cytomat Incubation Series provides robot-friendly random access storage of up to 1512 standard microplates, and can be easily integrated into any automated system. The series supplies an ideal environment for storage and incubation of assays in a temperature range of ambient +10°C up to 70°C (model dependent), with high humidity up to 95% r.H. and stable CO2 conditions.
Cytomat incubator series
Provides an ideal safe environment for storage and incubation of assays in a temperature range of ambient +10° to 50°C, with high humidity up to 95% r.H. stable CO2 conditions and automated decontamination routine ContraCon without harmful gases.
A midsize automated incubator for up to 105 (126) standard microplates, with a temperature range of ambient +10°C to 50°C, controllable humidity to 98% r.H., and stable CO2 conditions.
Fast, high-capacity cell growth and assay incubation with dependable sample protection for up to 210 standard microplates, a temperature range of ambient +10°C to 50°C combined with controlled humidity to 95% r.H, and stable CO2 conditions.
Ideal for highest capacity storage and incubation for up to 504 standard microplates, a temperature range of ambient +10°C to 50°C, humidity to 95% r.H., and stable CO2 conditions.
Two Cytomat 24 C incubators combined into one to accommodate up to 1008 standard microplates, a temperature range of ambient +10°C to 50°C, humidity to 95% r.H., and stable CO2 conditions.
Configuration options
The Cytomat system delivers the microplate to a single point, offering “easy-teach” integration for any automated system. A single loading and unloading point – the transfer station – makes teaching simple and cuts down the programming time, making for an extremely robot-friendly design.
These transfer stations offer a versatile placement of the Cytomat in the lab.
Standard configuration
Standard Transfer Station with Swap Station
Standard transfer station
This standard transfer station supplies the microplate in portrait format. A 90° turnable version is available for landscape presentation. 
Linear configuration
Linear Transfer Station and Plate Lift
Linear transfer station
This linear transfer station is available in different lengths (300mm, 400mm, 500mm, and customized lengths) for extended reach and delivers the microplate in portrait format. Additional options are available for landscape presentation and lateral movement.
Swap station (180°)
This swap station accelerates your lab automation handling system by providing two plate positions.
Plate lift
This plate lift is stepper motor driven (SMD) and offers microplate movement to a predefined height within a 500 mm stroke. 
The right options to match your application and automation needs with internal barcode readers, stackers, copper chambers and more. 
Internal Barcode Reader
Internal barcode reader
The barcode reader is located on the plate handler, allowing retrieval of information on every position, even when manually loaded.
Microplate Stacker
Stacker
The stainless steel, autoclavable Cytomat Stacker accommodates all standard microplates. Dedicated pitch sizes are available to optimize capacity for different types of microplates.
Copper Chamber
Copper chamber
Solid pure antimicrobial copper chambers are proven to be effective against contamination.
Additional options:
  • O2-control
  • hH – High humidity up to 98% at 37°C
  • Door left hinges (DL)
  • H2O2 generator ports
  • SiLA interface option
Many more options available
The Cytomat system delivers the microplate to a single point, offering “easy-teach” integration for any automated system. A single loading and unloading point – the transfer station – makes teaching simple and cuts down the programming time, making for an extremely robot-friendly design.
These transfer stations offer a versatile placement of the Cytomat in the lab.
Standard configuration
Standard Transfer Station with Swap Station
Standard transfer station
This standard transfer station supplies the microplate in portrait format. A 90° turnable version is available for landscape presentation. 
Linear configuration
Linear Transfer Station and Plate Lift
Linear transfer station
This linear transfer station is available in different lengths (300mm, 400mm, 500mm, and customized lengths) for extended reach and delivers the microplate in portrait format. Additional options are available for landscape presentation and lateral movement.
Swap station (180°)
This swap station accelerates your lab automation handling system by providing two plate positions.
Plate lift
This plate lift is stepper motor driven (SMD) and offers microplate movement to a predefined height within a 500 mm stroke. 
The right options to match your application and automation needs with internal barcode readers, stackers, copper chambers and more. 
Internal Barcode Reader
Internal barcode reader
The barcode reader is located on the plate handler, allowing retrieval of information on every position, even when manually loaded.
Microplate Stacker
Stacker
The stainless steel, autoclavable Cytomat Stacker accommodates all standard microplates. Dedicated pitch sizes are available to optimize capacity for different types of microplates.
Copper Chamber
Copper chamber
Solid pure antimicrobial copper chambers are proven to be effective against contamination.
Additional options:
  • O2-control
  • hH – High humidity up to 98% at 37°C
  • Door left hinges (DL)
  • H2O2 generator ports
  • SiLA interface option
Many more options available
Featured videos
Thermo Scientific Cytomat 10 Series
This video shows the Cytomat 10 automated incubator series combines superior environmental control, fast access times and versatility, making it the ideal choice for cell-based workflows requiring larger capacity incubation.
Video Player is loading.
Current Time 0:00
Duration 2:57
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:57
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextThermo Scientific Cytomat 10 Series
        This video shows the Cytomat 10 automated incubator series combines superior environmental control, fast access times and versatility, making it the ideal choice for cell-based workflows requiring larger capacity incubation.
      2. Now Playing
        Up NextThermo Scientific™ Cytomat™ 2 C-LiN Series Automated Incubator and Storage Devices
        The next generation of advanced incubation and storage solutions for cell culture applications.