Reliable microcentrifuges for everyday laboratory use
Thermo Scientific microcentrifuges perform a variety of microvolume protocols with fast acceleration and deceleration. These centrifuges offer many options, including a choice of two performance levels, ventilated or refrigerated models, and optional rotors to fit your application. Find the ideal blend of capabilities to support basic microtube processing, nucleic acid mini-preps, spin-columns, and hematocrit tests.
Key product highlights
Ease of use
Simplify operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure.
Fast and efficient
Fast acceleration and deceleration, and outstanding corrosion resistance with lightweight engineered polymer rotors; includes a broad array of microcentrifuge rotors and accessories for processing micro-volumes.
Protective of user and samples
Ergonomic one-click biocontainment lid opening and closing, as well as transparent design for added safety with certified1 ClickSeal biocontainment lids.
Small samples, fast processing
The 21,100 x g microcentrifuge models are designed for high-end research applications that demand the fastest runs, and the 17,000 x g models are excellent for routine lab work. All microcentrifuges feature intuitive controls, easy-to-read displays, and one-click lid closure.
Two standard rotors
Two standard rotors, made of lightweight engineered polymer to resist corrosion, are included with microcentrifuges and accept 1.5, 2, and 5 mL microtubes. Other polymer rotors support 0.2 and 0.5 mL microtubes, PCR strip tubes, and hematocrit capillary tubes. Some rotors accommodate ClickSeal biocontainment lids for added sample protection.
Available microcentrifuge models
Sorvall brand
Pico / Fresco brand
MicroCL brand
  • Pico 17 
    (ventilated, up to 17,000 x g)
  • Fresco 17 
    (refrigerated, up to 17,000 x g)
  • Pico 21 
    (ventilated, up to 21,100 x g)
  • Fresco 21 
    (refrigerated, up to 21,100 x g)
Product video and literature for microcentrifuges
Video Player is loading.
Current Time 0:00
Duration 1:21
Loaded: 0%
Stream Type LIVE
Remaining Time 1:21
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    1 Certified by the National Institute for Health Protection, Porton Down, UK or TÜV NORD CERT GmbH, Germany
    Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.