Specialized agitation without aggravation
 With precision performance and robust construction, our shakers offer a variety of controlled agitation types, including shaking, rocking, waving rotation, touch vortex mixing, and more. No matter how specific your applications needs are, we offer a specialized shaker that reliably performs to your specifications, while offering versatility and durability as your needs change.
Specific motion
Different applications require specific types of agitation, speed and duration to achieve optimal results. Our portfolio of specialized shakers, rockers, rotators, mixers offers this finely tuned functionality, helping to ensure predictable workflows. A wide range of capabilities are available and typically defined by type of motion, speeds and sample container compatibility appropriate for your application.
Agitation type
Linear shaking
Circular shaking
Vortex mixing
Thermal vortex mixing
Rotation
Waving rotation
Rocking
Rolling
Description
“Back and forth” agitation on a horizontal plane
Agitation of the sample container around a circumference on a horizontal plane
Provides a thorough mixing or blending of solutions using a swirling motion
Vortex blending with the ability to control an elevated temperature
360° orbital rotation for gentle incorporation on horizontal plane
360° orbital rotation along with planar rocking Gentle side-to-side planar rocking motion
Gentle back-and-forth rolling of cylindrical containers
Speed ranges
150 to 1,200rpm 100 to 8,500rpm 200 to 3000rpm
250 to 1,400rpm
5 to 500rpm 5 to 120rpm 1 to 70rpm 1 to 80rpm
Vessel compatibility
  • Microplates
  • Tubes
  • Microplates
  • Tubes
  • Microplates
  • Tubes
  • Flasks
  • Microplates
  • Microtube racks
  • Plates
  • Dishes
  • Culture bottles
  • Bags
  • Plates
  • Dishes
  • Culture bottles
  • Bags
  • Plates
  • Dishes
  • Tubes
  • Culture bottles
  • Bags
  •  
  • Tubes
  • Round bottles
Typical applications
  • PCR
  • Hybridization
  • Binding
  • Immunoassays
  • Bead assays
  • Buffers
  • Media
  • Circulated
  • Clinical
  • Blood -Chemistry
  • DNA extraction
  • Molecular biology
  • Food and beverage
  • Academia
  • Enzyme studies
  • Protein digestion
  • Genetic analysis
  • Metabolite studies
  • Molecular biology
  • Clinical biochemistry
  • Genetics
  • Genomics
  • Staining gels
  • ELISA
  • Antibody staining
  • Cell culture media
  • Bioproduction
Durable construction
Thermo Scientific specialized shakers are built to last. Our emphasis on quality manufacturing practices ensures that only the highest-grade raw materials are used, including high-strength aluminum, stainless steel hardware, and high-quality resins for plastic moldings. Our specialized shakers are certified under both cCSAus and CE.
Simplified, robust designs include durable, impact resistant shaker housing, thick rubberized feet for stability, durable, low or no-maintenance drive mechanisms and brushless motors to provide years of worry-free use.
2-year warranty
All of our specialized shakers include a 2-year warranty parts and labor, or product replacement warranty, based on service part availability.
Reliable performance
Thermal Mixer
We understand the strict performance parameters that are required for reproducible results. Our shakers and mixers are engineered for reliable performance and durability while meeting your specific sample processing needs. Speed, time and even angle adjustments offer precision with the adaptability to use in a wide variety of assays, optimizing your valuable bench space.
Digital display and simple controls
Newer models, such as the Thermal Mixer, are equipped with upgraded, digital LCD displays for at-a-glance shaking data. All models are designed with simple touch pad controls for speed, timing and temperature (for Thermal control models).
Accurate speed controls
Easily set shaking speed with touch pad controls and be assured that the shaker will perform within 1 –2 RPM depending on speed setting. Preset options allow you to quickly achieve your desired motion for more accuracy.
Adjustable angles
Models that operate on angles (e.g., rotators and rockers), include planar angle adjustability. These angle adjustments are easy to use and vary from 95° to 180°. These wide-angle ranges can help maintain the suspension state of biological samples such as blood mixing.
Ease of use
Our benchtop agitators and shakers of all types are designed to be precise and intuitive. Touch pad controls offer quick set up for time, speed, and temperature-specific operations. Preset options allow for immediate and repetitive use without set up. And because of the simplified and durable construction including spill resistant and sealed exteriors, making cleaning and maintenance easy.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Touch mode and pre-set options for fast and repetitive use
      Models designed for routine mixing operations, such as the Digital Vortex Mixer, offer touch mode and pre-set options that allow user to simply hold the sample on the platform for a quick mix.
      Automation Enabled
      Thermal Mixer Automation
      Thermo Scientific Magnetic Microplate Shakers were developed automation processing and SBS microplates of all sizes. Supporting a range of versatile applications, our Magnetic Microplate Shakers are designed for consistent circular and linear agitation that supports high-speed integrated applications. These shakers allow for simultaneous control of up to 14 shakers through master system software and raise and lower microplates for use with robotic workstations.
      For Research Use Only. Not for use in diagnostic procedures.