Meet ICH Q1B guidelines with temperature, humidity and light control
These stability chambers provide precise control over a wide range of temperature and humidity settings just like our other stability chambers, but also include lighting modules for light stability testing, plant and insect growth studies. They are ideal for product stability studies, shelf-life testing and other large-scale biological research.
Key features
Flexible lighting
Variable set-up addresses requirements for sample distance to light and sample volume.
ICH light module
Optimal light exposure according to guideline Q1B.
Biological lighting
Specific light modules for plant growth and animal hatching available.
Solid door
Ensures there is no "light pollution" in the light exposure to samples.
Wide light distribution
Achieved with 7 fluorescent light bulbs.
Controlling relative humidity in vertical light stability chambers
This chart illustrates the maximum and minimum attainable humidity within the vertical light stability chambers at a given set temperature with the light on and two light modules installed. Optional heatless dryer is available for lower humidity levels.
Forma Environmental Chamber humidity range
Specifications
Large Vertical Light
Stability Chamber
Small Vertical Light
Stability Chamber
Size
29 cu. ft.
11 cu. ft.
Exterior (W x H x D)
965 x 2248 x 813mm
38.00 x 88.50 x 32.00 in.
965 x 1308 x 813mm
38.00 x 51.50 x 32.00 in.
Interior (W x H x D)
787 x 1524 x 686mm
31.00 x 60.00 x 27.00 in
787 x 609 x 686mm
31.00 x 24.00 x 27.00 in.
Net weight
347kg / 765 lbs
261kg / 575 lbs
Shipping weight
472kg / 1040 lbs
325kg / 715 lbs
Set Light Temperature – No Humidity
Max # of light modules
3
2
Temperature range with light on
25°C to 50°C (68°F to 122°F)
10°C to 50°C (50°F to 122°F)
Controller, sensor
Watlow Microprocessor - PID, with RTD sensor
Display
Digital dual LED, 0.1°C steps readability & setability
Programmability
Single set point, ramp & soak (to 40 steps)
Temp. uniformity @ 20 to 37°C
≤ ±3.0°C
Temp. stability @ 20 to 37°C
≤ ±3.0°C
Set Light Temperature and Humidity
Max # of light modules
2
Temperature range
15°C to 50°C (59°F to 122°F)
10°C to 50°C (50°F to 122°F)
Temp. uniformity @ 20 to 40°C
≤ ±2.5 °C
≤ ±2.3 °C
Temp. stability @ 30°C, 75% RH
≤ ±0.5°C
Water consumption / specification
Max. 12L per day / 3 gal per day;
demineralized water, resistance: 0.5 - 1 MΩ cm
Relative humidity range
Above ambient to 75% @ 50°C (122°F) 
Humidity stability
≤ ±5% RH
Water connection*
Fill port: 1⁄8" female NPT, drain port: 3⁄8" FPT & 3⁄8" O.D. P-trap right rear, initial fill approx. 1L / 1qt.
Electrical
Voltage
208-220V, 60Hz
Plug
NEMA 14-20P
Power switch
2-Pole
Dry Contacts
Common, NO, NC
Data Output
4-20 milliamp, temperature and RH
Access port
54mm / 2.1in diameter; left side; with exterior and interior silicone stopper
Options
Cabinet Options Available
Choice of access ports
Duplex sockets
Choice of chart recorders
Infrared CO2 Control Package
Choice of door types and styles, including Lexan(tm) inner doors
Choice of shaker support systems
Temperature mapping and calibration certificate
Humidity Systems Options Available
Heatless dryer
Condensate pump kit
Condensate evaporator
Carboy kit
Order Number
3942
3909
Choose light modules for ideal for ICH Q1B light stability testing, plant growth or animal/insect hatching
The light modules for plant growth emit increased red and blue wavelengths based on typical sunlight patterns.  Light modules for insect hatching mimic daylight – ideal for the required day/night simulation – including dawn and dusk progression settings. The light modules for light stability testing accommodate ICH guidelines Q1B and have a combination of bulbs that address the required wavelengths, and UV light according to these guidelines.