Pipette Tips
Tips for any pipette - micropipette, filter tips, low retention and more
Accurate and precise liquid measurement is critical to the success of your research and selecting the right pipette tip for your pipette will improve performance and your results.
We make it easy to find and select the pipette tips you need to deliver consistency and reliability every time you pipette, whether you have a Thermo Scientific Finnpipette or ClipTip pipetting system or you are looking for a universal pipette tip, for whatever pipette you are using.
InVitro Diagnostic Regulation Update
Choose the tips that are right for you
ART barrier tips
  • Self-sealing barrier to completely block contamination
  • Universal fit for virtually any brand of pipette
  • Low retention for optimal transference
  • ART Barrier or non-filtered available
    View selection tool ›
Finntip, Finntip Flex Tips
ClipTip Tips
  • Interlocking technology for secure seal that won’t loosen or fall off
  • Minimal attachment and ejection forces
    View selection tool ›
SoftFitL-family
  • Designed for Rainin™ LTS-style pipettes
  • Positive stop design maintains ergonomic benefits of pipette
    View selection tool ›
Thermo Fisher Scientific has been providing innovative liquid handling solutions to laboratories like yours for decades. Rely on our expertise and innovation for all your pipetting needs.
Find the pipette tips with the features you need
 ART UniversalFinntipFinntip Flex ClipTip
Lower attachment
and ejection forces
System tip provides optimal performance with companion pipette 
Universal fit for most
brands of pipettes
Self-sealing barrier for 100% contamination prevention   
Color-coded volume identification
Eco-friendly packaging options
384-format available  
Pipette and pipette tips compatibility
 ART UniversalFinntipFinntip Flex ClipTip
Finnpipette F1 and
Finnpipette F2
Finnpipette Novus 
F1-ClipTip   
E1-ClipTip   
Gilson™ pipettes   
Eppendorf™ pipettes   
Rainin™ Classic pipettes*   
Other pipette brands   
 Learn moreLearn moreLearn moreLearn more
*For Rainin™ LTS pipettes, see SoftFit-L tips
GLP guide
A comprehensive Good Laboratory Pipetting (GLP) guide to help you achieve better results by providing tips on advancing your skills.
Download the guide now ›
ART SoftFit-L tip
ART-SoftFit-L-hinged-rack
SoftFit-L Tips
The SoftFit-L Tip was designed for optimal performance with Rainin™ LTS pipettes. SoftFit-L Tips deliver the accuracy and precision you expect and maintain the ergonomic benefits of the pipette with a positive-stop collar that reduces tip attachment and ejection forces.
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.