Build a healthier future with trusted and proven microarray solutions
Leading labs like yours can advance carrier screening research with our comprehensive genetic analysis software with intuitive and flexible workflows to help improve efficiencies and implement a scalable solution.
Workflow
Thermo Fisher Scientific offers powerful microarray solutions that are powered with robust workflows that make it easy to obtain consistent, reliable, and high-quality results critical to carrier screening research.
Our complete microarray platform includes assays, automated and manual target preparation options, fully kitted reagents, an array processing instrument, and data analysis software for simplified interpretation and reporting. These tools support research laboratories to help maximize operational time.
Image of Carrier Screening workflow
Product overview
Reproductive Health Analysis Software
Reproductive Health Analysis Software (RHAS) for cytogenetic and variant analysis enables visualization and summarization of chromosomal aberrations, genotyping of specified variants, as well as SMN1 research.
RHAS provides tools to:
  • Perform genotyping of CEL files from CarrierScan assay plates
  • Run QC, copy number, genotyping, variant determination, and SMN1 study algorithms in selected workflows
  • View sample QC data in tables and graphs
  • View variant data in cluster plots for all probe sets
  • View variant data in tables or as a batch summary
  • Export your data in various formats for use in third-party software
  • View SMN study results in table and graphical formats
Image of Software screenshot MSV Window
MSV window
Variant table cluster plot 
Carrier Reporter Software
Our cloud-based, fit-for-purpose interpretation and reporting research software is powered by Igentify™ and follows the American College of Medical Genetics and Genomics (ACMG) tiers 1-3 and provides coverage of >600 of autosomal, X-linked recessive diseases. It is capable of analyzing tens of thousands of sequence variants, including those involved in spinal muscular atrophy (SMA), Duchenne muscular dystrophy (DMD), cystic fibrosis, sickle cell disease, Tay-Sachs disease, and hemoglobinopathies.
Carrier Reporter Software also enables research labs to consolidate their microarray results with SMN1/2 and FMR results from the CarrierMax Reporter Software. 
Carrier reporter software
Get a holistic view into queued sample results, assign to individuals, or analysis of your assigned samples
Image of Software screenshot of variant info
User-friendly interface for the interpretation and analysis of variants
Image of Software screenshot of single or paired samples
Quickly and easily generate intuitive results reporting for either a single sample or paired samples
CarrierMax Reporter Software
CarrierMax Reporter Software supports analysis for the CarrierMax FMR1 and SMN1/2 assays. The software is used in conjunction with GeneMapper Software, which is a flexible fragment analysis software package that provides quality DNA sizing and allele calls for all Applied Biosystems genetic analyzers. This software is designed to support multi-application functionality, including analysis of amplified fragment length polymorphism (AFLP), loss of heterozygosity (LOH), microsatellites, and SNP genotyping.
CarrierMax software capabilities include:
  • Summary of samples analyzed
  • Translation of selected GeneMapper software peaks to CGG repeats
  • Consolidation of CGG repeat alleles identified  
  • Final FMR1 classification
  • Translation of selected GeneMapper Software peaks to SMN1 and SMN2 exon 7 copy number
  • Report of detected 2 + 0 associated SNPs
  • Final SMN1 exon 7 carrier classification
  • Results export for interpretation and reporting
For Research Use Only. Not for use in diagnostic procedures.