Search Thermo Fisher Scientific
The emergence of the Human Genome Project equipped researchers with new molecular tools for gene discovery and gene expression. It enabled researchers to identify the genetic defects associated with complex diseases, including common neurological diseases, and ushered in a new era of analysis.
Complex diseases, such as Parkinson’s and Alzheimer’s Disease, are caused by a combination of genetic and environmental factors. At the heart of understanding and thereby treating complex diseases is identifying the many factors, both genetic and environmental, and the interaction of these factors, that affect disease onset and progression.
TaqMan Gene Expression Assays allow researchers to use real-time PCR to investigate how gene expression is changed in samples taken from an individual with a complex disease. Furthermore, TaqMan Assays are useful in in vitro studies focusing on a single genetic or environmental factor thought to be in involved in complex disease.
Download a list of genes and corresponding Assay IDs for the most popular biomarkers associated with complex and neurological diseases.
Search for a predesigned TaqMan Assay using the gene symbol in the tool below.
What chromosome position are you interested in?
Genome Version: Build GRCh38 for
In addition to single tube assays, we also have a variety of both configurable and fixed content TaqMan Array products focused on complex disease research. TaqMan Arrays are pre-spotted 96-well plates or 384-well microfluidic array cards, saving you the time of pipetting many assays into an empty plate. Just add your cDNA and master mix and you’re ready to go!
TaqMan Array Human Alzheimer's Disease
96-well (0.2 mL) plate format containing 92 gene targets and 4 endogenous controls
TaqMan Array Human Alzheimer's Disease, Fast
96-well (0.1 mL) format containing the same set of 96 targets
TaqMan Array Human Alzheimer's Panel
384-well array card format; test 96 targets in four samples in a single experiment with the efficient workflow advantage of TaqMan Array Cards; fill 384 wells in less than 15 minutes
TaqMan Array Mouse Alzheimer's Disease
96-well (0.2 mL) plate format containing 92 gene targets and four endogenous controls
TaqMan Array Mouse Alzheimer's Disease, Fast
96-well (0.1 mL) format
TaqMan Array Mouse Alzheimer's Panel
384-well array card format
TaqMan Array Human Huntington’s Disease
96-well (0.2 mL) plate format containing 92 gene targets and four endogenous controls
TaqMan Array Human Huntington’s Disease, Fast
96-well (0.1 mL) format
TaqMan Array Human Pathogenesis of Rheumatoid Arthritis
96-well (0.2 mL) plate; test 48 targets in two samples in a single plate
TaqMan Array Human Pathogenesis of Rheumatoid Arthritis, Fast
96-well (0.1 mL) format
TaqMan Array Human Pathogenesis of ALS
96-well (0.2 mL) plate; test 32 targets in three samples in a single plate
TaqMan Array Human Pathogenesis of ALS, Fast
96-well (0.1 mL) format
For Research Use Only. Not for use in diagnostic procedures.