Search Thermo Fisher Scientific
Predesigned TaqMan™ Copy Number Assays are ideal for analysis of copy number variation (CNV) and smaller regions in human and mouse genomes. CNV is an important polymorphism associated with diseases such as cancer, immune diseases and neurological disorders as well as drug metabolism.
The human assay collection includes over 1.6 million assays targeting gene exons and introns, extragenic regions, and CNV sequences from the Database of Genomic Variants (DGV). The mouse collection includes over 180,000 assays targeting gene exons. Assays to common vector marker and reporter genes are also available for transgenic studies.
What chromosome position are you interested in?
Genome Version: Build GRCh38 for
Need a custom designed TaqMan Copy Number Assay? Start here ›
Pre-formulated Assay |
|
Documentation | Available to download at thermofisher.com/taqmanfiles :
|
Assay Information File (AIF) | Predesigned Assay AIF includes: sales order number, ship date, part number, product type, assay ID, lot number, assay mix concentration, primer & probe concentrations, reporter dye & quencher, context sequence (includes probe region), gene ID (if applicable), DGV ID (if applicable), species, cytogenetic band, location on NCBI reference genome assembly (middle of context sequence), assembly build version, and other information. Assay sequences are not provided. |
Reactions | Sufficient for 360, 750, or 2,900 20-uL reactions in a 96-well plate, or 720, 1,500 or 5,800 10-uL reactions in a 384-well plate (depending on scale ordered) |
Tracking/Identification |
|
Expiration Date | 5 years from date of manufacture. |
Shipping and Storage | Shipped at ambient temperature. |
Catalog # | Size | # of 20 µL Reactions (96-well) | # of 10 µL Reactions (384-well) |
---|---|---|---|
4400291 | S: 20X concentration | 360 | 720 |
4400292 | M: 20X concentration | 750 | 1,500 |
4400293 | L: 60X concentration | 2,900 | 5,800 |
Approximate delivery times are 4-6 days in the US and 6-10 days in Europe.
Protocol: TaqMan® Copy Number Assays
Quick Reference Card: CopyCaller™ Software
User Guide: CopyCaller™ Software
Fact Sheet: TaqMan® Assay ambient shipping
White Paper: TaqMan® Assays ambient shipping
Application Note: Design Pipeline and Validation of TaqMan® Copy Number Assays
Product Bulletin: TaqMan® Copy Number Assays
Product Overview: TaqMan® Copy Number Assays
Poster: Coverage of the PharmaADME Consortium Core Marker List by TaqMan® DME and Copy Number Assays
Experiment Workflow for Copy Number Variation Analysis using Real-Time PCR
Copy Number Variation Analysis Software
Real-Time PCR Troubleshooting Tool
Low copy number of the salivary amylase gene predisposes to obesity.
VOLUME 46. NUMBER 5. MAY 2014 Nature Genetics. Mario Falchi et. al.
Pre-formulated Assay |
|
Documentation | Available to download at thermofisher.com/taqmanfiles :
|
Assay Information File (AIF) | Predesigned Assay AIF includes: sales order number, ship date, part number, product type, assay ID, lot number, assay mix concentration, primer & probe concentrations, reporter dye & quencher, context sequence (includes probe region), gene ID (if applicable), DGV ID (if applicable), species, cytogenetic band, location on NCBI reference genome assembly (middle of context sequence), assembly build version, and other information. Assay sequences are not provided. |
Reactions | Sufficient for 360, 750, or 2,900 20-uL reactions in a 96-well plate, or 720, 1,500 or 5,800 10-uL reactions in a 384-well plate (depending on scale ordered) |
Tracking/Identification |
|
Expiration Date | 5 years from date of manufacture. |
Shipping and Storage | Shipped at ambient temperature. |
Catalog # | Size | # of 20 µL Reactions (96-well) | # of 10 µL Reactions (384-well) |
---|---|---|---|
4400291 | S: 20X concentration | 360 | 720 |
4400292 | M: 20X concentration | 750 | 1,500 |
4400293 | L: 60X concentration | 2,900 | 5,800 |
Approximate delivery times are 4-6 days in the US and 6-10 days in Europe.
Protocol: TaqMan® Copy Number Assays
Quick Reference Card: CopyCaller™ Software
User Guide: CopyCaller™ Software
Fact Sheet: TaqMan® Assay ambient shipping
White Paper: TaqMan® Assays ambient shipping
Application Note: Design Pipeline and Validation of TaqMan® Copy Number Assays
Product Bulletin: TaqMan® Copy Number Assays
Product Overview: TaqMan® Copy Number Assays
Poster: Coverage of the PharmaADME Consortium Core Marker List by TaqMan® DME and Copy Number Assays
Experiment Workflow for Copy Number Variation Analysis using Real-Time PCR
Copy Number Variation Analysis Software
Real-Time PCR Troubleshooting Tool
Low copy number of the salivary amylase gene predisposes to obesity.
VOLUME 46. NUMBER 5. MAY 2014 Nature Genetics. Mario Falchi et. al.
For Research Use Only. Not for use in diagnostic procedures.