Detect your target by film, CCD cameras or laser-based imagers
by Thermo Scientific Pierce Protein Biology Team - 06/01/11
The Thermo Scientific Pierce ECL Plus Substrate is a chemiluminescent and chemifluorescent peroxidase substrate for western blotting. The Pierce ECL Plus Substrate generates acridinium esters when it reacts with horseradish peroxidase (HRP), producing a strong signal that is equal to or better than similar commercially available substrates (Figure 1). The ester intermediates react with peroxide and produce a strong and sustained chemiluminescent signal that can be captured by X-ray film or a CCD camera. A robust fluorescent signal is also produced, which can be captured with fluorescence imagers (Figure 2).
32132-v15-Fig1-ECL-Plus
Figure 1. Thermo Scientific Pierce ECL Plus Substrate performs as well or better than substrates from other suppliers. HeLa cell lysate was diluted in SDS-PAGE sample buffer and heated to 95˚C for 5 minutes. Lane 1 contained 10μg of total protein. Four 1:1 dilutions were prepared and applied to Lanes 2-5 at 10μL/well. The proteins were transferred to nitrocellulose membranes (Part No. 88013). The membranes were blocked with 5% milk in Tris-buffered saline with Tween*-20 Detergent. Rabbit anti-hsp86 antibody (Ab No. PA3-013) diluted 1:1000 and goat anti-rabbit HRP (Part No. 31460) at 6.6ng/mL were used for target detection. Signal was detected using the indicated substrates and Thermo Scientific CL-XPosure Film (Part No. 34090).
32132-v15-Fig2-ECL-Plus
Figure 2. One substrate detected by three methods. Blots were prepared as described in Figure 1. The target was detected with rabbit anti-MAP kinase (EMD Millipore) diluted 1:1000, goat anti-rabbit HRP (Part No. 31460) at 6.6ng/mL and Pierce ECL Plus Substrate. The membranes were exposed to CL-XPosure Film for five seconds and scanned using the Typhoon* 9410 Variable Mode Imager (excitation at 457nm, emission at 510nm) and Syngene* G:Box iChemiXT Imager (1 minute exposure).
Pierce ECL Plus Substrate enables detection of low-picogram amounts of target protein (Figure 3) on nitrocellulose and PVDF membranes when probed with the appropriate primary and secondary antibody concentrations. This versatile and reliable western blotting substrate has a broad dynamic range and excellent sensitivity combined with a long-lasting signal (Figure 4).
32132-v15-Fig3-ECL-Plus
Figure 3. Chemiluminescent signal can be detected for hours after substrate incubation. Blots were prepared as described in Figure 1. The target was detected with rabbit anti-hsp86 (Ab No. PA3-013) diluted 1:1000 and goat anti-rabbit HRP (Part No. 31460) at 6.6ng/mL. Signal was detected with CL-XPosure Film exposed at the indicated times.
32132-v15-Fig4-ECL-Plus
Figure 4. Detect low-picogram amounts of target protein. GST (500ng) was loaded onto Lane 1 and serially diluted 1:1 and loaded onto Lanes 2-9 (10μL/well). Proteins were transferred to nitrocellulose membrane (Part No. 88013) and Thermo Scientific SuperBlock Blocking Buffer TBS (Part No. 37535) was used to block nonspecific binding sites. GST was detected with biotinylated anti-GST (Santa Cruz) at 0.5μg/mL and Thermo Scientific Pierce High Sensitivity Streptavidin-HRP (Part No. 21130) diluted 1:60,000. Signal was detected with CL-XPosure Film exposed for 5 seconds.
Pierce™ ECL Plus Western Blotting Substrate
Thermo Scientific Pierce ECL Plus Substrate is an acridan-based chemiluminescent and chemifluorescent HRP substrate for Western blot detection using X-ray film or CCD- or laser-based imagers. This product is sold as Pierce ECL 2 Substrate (Cat. #PI80196) through the Fisher Scientific channel and other channels.
Features of ECL Plus Substrate:
  • Easy to use—can be substituted for the discontinued GE ECL Plus Substrate without any re-optimization
  • Higher sensitivity—detect targets down to the low-picogram level
  • Longer signal duration—sustained light output for as long as 5 hours
  • More imaging options—X-ray, CCD or laser-based imagers
  • More affordable—high quality and performance at a lower price than other suppliers' similar substrates
Learn more about Thermo Scientific Pierce ECL Plus Substrate
Related products
Catalog # Name Size Price (USD) Qty
88013 Nitrocellulose Membranes, 0.2 μm, 15 Pre-cut Membranes Each
163.65
Online Exclusive
184.00
Save 20.35 (11%)
21130 Pierce™ High Sensitivity Streptavidin-HRP, 0.5 mL Each
263.65
Online Exclusive
296.00
Save 32.35 (11%)
37535 SuperBlock™ (TBS) Blocking Buffer, 1 L Each
243.65
Online Exclusive
273.00
Save 29.35 (11%)
31460 Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP 2 mL
259.00
34090 CL-XPosure™ Film 100 sheets
215.65
Online Exclusive
228.00
Save 12.35 (5%)
For Research Use Only. Not for use in diagnostic procedures.