promo-pod-protein-230x195
The Flp-In™ System allows for stable integration and expression of your gene of interest to deliver single-copy isogenic cell lines. Flp-In™ expression involves introduction of a Flp Recombination Target (FRT) site into the genome of the mammalian cell line of choice. An expression vector containing your gene of interest is then integrated into the genome via Flp recombinase–mediated DNA recombination at the FRT site.
We offer a selection of Flp-In™ products, including expression vectors and systems, as well as parental cell lines with a stably integrated FRT site. Designed for rapid generation of stable cell lines, these products ensure high-level expression of your protein of interest from a Flp-In™ expression vector.
Protein vector selection tool
Ordering Information
Catalog # Name Size Price (USD) Qty
R78007 Flp-In™ T-REx™ 293 Cell Line Each
3,125.00
R75807 Flp-In™-CHO Cell Line Each
2,571.65
Online Exclusive
2,795.00
Save 223.35 (8%)
R75007 Flp-In™-293 Cell Line Each
2,696.65
Online Exclusive
2,970.00
Save 273.35 (9%)
R76207 Flp-In™-Jurkat Cell Line Each
2,795.00
R76107 Flp-In™-3T3 Cell Line Each
2,800.00
K601001 Flp-In™ Complete System Each
2,580.00
K650001 Flp-In™ T-REx™ Core Kit Each
2,758.65
Online Exclusive
2,950.00
Save 191.35 (6%)
K601002 Flp-In™ Core System Each
1,866.00
K602001 pcDNA™5/FRT/V5-His-TOPO™ TA Expression Kit 20 reactions -
V652020 pcDNA™5/FRT/TO Vector Kit 20 µg
1,232.00
V601020 pcDNA™5/FRT Mammalian Expression Vector 20 µg
1,080.00
K602501 pSecTag/FRT/V5-His© TOPO™ TA Expression Kit 20 reactions -
K603501 pEF5/FRT/V5 Directional TOPO™ Cloning Kit 20 reactions -
V600520 pOG44 Flp-Recombinase Expression Vector 20 µg
916.00
V601520 pFRT/lacZeo Vector 20 µg
816.00
V602020 pEF5/FRT/V5-DEST™ Gateway™ Vector 6 µg
876.00
For Research Use Only. Not for use in diagnostic procedures.