protein-expression-mondrian
We offer a range of protein expression systems and tools that are easy to use, designed to deliver high protein yields for a wide variety of downstream applications, and allow you to achieve results quickly. Creating your gene, purifying your plasmid, and selecting suitable protein expression products are simplified with help from our selection tools and educational videos.
Protein expression workflow
Protein expression workflowgeneart-assemblygeneart-synthesisoneshotbenchpropurelinkexpi293lipopichiabacecoli
Selecting a host system for protein expression & production
When selecting a host system for expressing protein there are many factors to consider. These include: post translational modifications, equipment availability, and time constraints. We have mapped out the details for each host system, that along with information for applicable products the we offer, to allow you to make the most appropriate choice based on your needs and budget.
 Do you need modifications?How quickly do you require protein?What capabilities do you have to support expression (lab equipment & resources)?
 NoneSomeCompleteDaysWeeksMonthsNoneSomeMany
Mammalian         
Bacteria         
Yeast         
Insect         
Get more detail about comparing protein expression systems
Equipment
  • What equipment is available for use?
Type of protein
  • Is your protein endogenously expressed in bacterial or yeast cells?
  • Do you know that it’s insoluble in a bacterial expression system?
  • Do you need native posttranslation modifications?
  • Do you want some level of eukaryotic posttranslational modifications?
Desired preparation time
  • One week or less (bacterial, transient mammalian)
  • One month or less (in addition to bacterial and transient mammalian, yeast & insect systems can be used)
  • More than one month (in addition to the systems above, you can use stable mammalian systems)  
Mammalian expression
Vector selectionCloningTransform in E. coliPlasmid purificationTransfectionExpressionPurificationAnalysis
Determined by promoter and terminator
Choose now
Determined by vector choice
- or -
GeneArt® Gene Synthesis
Recommend DH5α™ TOP10 strain
Choose now
Could be maxi, mega, or giga
Choose now
Depends on speed
Choose now
Media based on expression system choice: 293 or FreeStyle™
Choose now
Affinity based with tag or native
Choose now
Analysis & Assay
Choose now
Bacterial expression
Vector selectionCloningTransform in E. coliPlasmid purificationTransformOptimization of cloneScale & purifyAnalysis
Any vector with a T7 promoter
Choose now
Choose any cloning method
- or -
GeneArt® Gene Synthesis
Recommend TOP10 strain but can choose from subcloning products
Choose now
Mini prep kits
Choose now
Transform into any expression strain
Choose now
TB or LB
Choose now
TB, LB, or MagicMedia™
Choose now
& resin purification
Choose now
Analysis & Assay
Choose now
Yeast expression
Vector selectionCloningTransform in E. coliPlasmid purificationTransform into yeastClone selectionExpressPurificationAnalysis
Does protein have a secretion signal
Choose now
Yes? Use PichiaPink™
Clone with restriction enzymes
Choose now
One Shot® TOP10 Electrocomp™ E. coli
Choose now
Midi or maxi prep depending on yield requirement
Yeast strain
Zeocin™
Choose now
Media
Choose now
With secretion sometimes not necessary & affinity for tag
Choose now
Analysis & Assay
Choose now
Insect expression
Vector selectionCloningTransform in E. coliPlasmid purificationTransform into BacPurify BacmidTransfectAmplify/ ExpressAnalysis
Choose TOPO® or Gateway® vector
Choose now
Select  TOPO® or Gateway® for cloning
Choose now
Recommend DH5α™ TOP10 strain but can choose from subcloning products
Choose now
Mini prep kits
Choose now
Transform into DH10Bac
Midi or maxi prep- depending on yield requirement
Transfection with Cellfectin® II
SF9 & SF21
Choose now
Analysis & Assay
Choose now
How-to videos
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      "Meet the Inventor" Series
      Clone: 20% Off GeneArt® Expression-Optimized Genes ›
      Improve protein expression up to 100-fold with GeneArt® expression-optimized genes.