Reliable, versatile equipment to help improve sample storage workflow
Thermo Scientific barcode readers, plate sealers and tube cappers/decappers are trusted in labs worldwide to speed processing times, enhance workflow efficiencies and help ensure sample integrity. Our equipment is designed to enable reliable performance, be compatible with a wide range of Thermo Scientific consumables, and to be scalable with your throughput needs, with ease of implementation and use. 
Calibrated to your throughput
Regardless of your lab’s sample throughput needs, our sample storage equipment can improve your workflow efficiency, speed and reliability. Our portfolio includes manually operated and automated equipment options appropriate to help enhance sample storage workflows whether your lab moves just a few or thousands of samples through the lab daily. 
Barcode Readers
Cappers/Decappers
Plate Sealers
Available Speed Ranges
Rapid single tube reads
As little as 1 second for full racks
4 seconds for 8 tubes
Entire rack in less than 60 seconds
10-15 seconds/plate
2-3 plates/minute
5-6 plates/minute
Operation Types Available
Handheld/Manual
Automated
Handheld/Manual
Hands free/Automated
Manual
Automated
Throughput Ranges Available
Low
Medium
High
3-5 Racks/Day
10+ Racks/Day
Low
Medium
High
Consumable Compatibility
Our portfolio of sample storage equipment is designed to be compatible with the most popular and widely used Thermo Scientific consumables. Thermo Scientific Barcode Readers accommodate 1D, 2D, linear and human readable codes on tubes and racks. Our capping/decapping equipment is compatible with a wide range of Nunc and Matrix tubes and racks. Thermo Scientific Plate Sealers support deepwell plates, micro plates, well blocks, and racked tubes, along with a host of seal types from heat and adhesive seals to lids and capmats. Flexibility is built into our equipment designs so that your individual lab processes and preferences are accommodated out of the box, including many integrated lab automation systems. 
2D barcoding available on cryogenic storage tubes
Increased Sample security
In addition to the obvious benefits of being able to work faster and more efficiently, barcode readers, plate sealers and capping/decapping equipment help provide increased security for your valuable samples.  Using capping equipment enables an optimal torque for tube sealing while helping to reduce the risk of damaging the seal integrity over repeated capping and decapping procedures. Similarly, plate sealers apply the proper pressure and uniformity of seal across the entirety of the plate and for the type of seal material you use. Barcode readers help reduce data errors and enable more assured tracking through processing and storage procedures. Our reliable sample storage equipment eases workflow and helps give you more confidence in sample integrity. 
Scientist inspects plates
Proper tube sealing torque is critical to sample security
Sample loss due to evaporation during long-term cryogenic storage is a common problem that can prove troublesome or even disastrous. Learn more by reviewing this study that demonstrates how the application torque applied to the tube closure during capping affects sample attrition following repeated thawing and freezing cycles.
weight lost by torque me a % of the original volume
Ease of Use
From handheld, manual equipment to fully integrated, lab automation components, our sample storage equipment is designed to be easy to use out of the box and reliable over the long term. Manual devices incorporate ergonomic grips and robust mechanical components to help reduce users’ physical strain and risk of repetitive stress injury.
Automated devices incorporate simple, clear user interfaces, touch controls and intuitive software that help get you up and running quickly and keep you there. Workflow flexibility is also built in, allowing for easy switching between consumable sizes and formats on the fly.
Scientist with automated equipment
More affordable automation
Our portfolio of sample storage equipment is designed to help provide labs with enhanced workflow efficiency.  By integrating with other equipment necessary for your workflow, labs can scale up automation at a pace and cost that is more approachable.
automation plates
For general laboratory use.