Click on the tabs below to discover how the lack of genomic integration and viral remnants allows the use of CytoTune in a broad range of experiments, including disease research.
CytoTune - EmGFP Sendai Fluorescence Reporter
Figure 1: Fluorescence microscopy images of green BJ HDFn cells on black background. Green expression demonstrates ptake of Sendai virus into the cells.
BJ HDFn cells transduced with CytoTune - EmGFP Sendai Fluorescence Reporter at an MOI of 5 shown at 48 hours.   Green expression demonstrates the uptake of Sendai virus into the cells.
Successful iPSC reprogramming from a range of somatic cell types
The following somatic cell types have been successfully reprogrammed into iPSCs using CytoTune technology.
  • Adult and neonatal dermal fibroblasts
  • Amniotic fluid MSCs
  • Cardiac fibroblasts
  • CD34+ blood cells
  • Mammary epithelial cells
  • Nasal epithelial cells
  • Peripheral blood mononuclear cells (PBMCs)
  • Skeletal myoblasts
  • T cells
  • Umbilical vein epithelial cells
  • Urine epithelial cells
View endpoint PCR & staining
Lack of genomic integration and viral remnants allows the use of the CytoTune-iPS Sendai Reprogramming Kit in a broad range of experiments, including disease research.
Comparison of the lifecycles of non-integrating Sendai vectors and integrating vectors
Figure 3A: Schematic shows upper left corner of a cell including the cytoplasm (surrounding tan outline) and the nucleus (circular blue middle). Arrows and icons demonstrate how Sendai virus is introduced to the cell and works within it. Figure 3B: Schematic shows upper right corner of a cell including the cytoplasm (surrounding tan outline) and the nucleus (circular blue middle). Arrows and icons demonstrate how other vectors (besides Sendai virus) are introduced to the cell and work within it.
Sendai Virus (SeV) is an enveloped virus of 150–250 nm in diameter whose genome is a single chain RNA in the minus sense. Six genes coding for viral proteins are situated sequentially on the genome of the wild-type SeV in the following order.
Nucleocapsid protein (NP) forms the core nucleocapsid complex with the genome RNA. Phosphoprotein (P) is the small subunit of the RNA polymerase.  Matrix protein (M) supports the envelope structure from the inside. Fusion protein (F) fuses the viral envelope with the cell membrane when the virus enters the cell. The gene encoding the F protein is deleted from the CytoTune® -iPS Sendai Reprogramming Kit vectors, rendering them incapable of producing infectious particles from infected cells. 
Hemagglutinin-Neuraminidase (HN) recognizes the cell surface receptor, sialic acid.  Large protein (L) is the large subunit of RNA polymerase.
Because SeV infects cells by attaching itself to the sialic acid receptor present on the surface of many different cells, it can infect a wide range of cell types of various animal species. Activation of F protein by a protease is required for the virus-cell fusion process to take place. After infection, the virus goes through genome replication and protein synthesis in the cytoplasm, and then daughter virus particles are assembled and released.
Other viral vectors, such as lentivirus or retrovirus, require integration into the host genome for replication and can therefore disrupt the genome of the cells that are reprogrammed rendering iPSCs and their derivatives less safe for clinical applications or altering gene expression patterns which can compromise compound screens or disease pathway analyses. A single transduction with the CytoTune® -iPS Sendai Reprogramming Kit generates iPSCs at high efficiency with no genomic integration or viral remnants allowing the use of iPSCs and their derivatives in a broader range of research experiments.
CytoTune - EmGFP Sendai Fluorescence Reporter
Figure 1: Fluorescence microscopy images of green BJ HDFn cells on black background. Green expression demonstrates ptake of Sendai virus into the cells.
BJ HDFn cells transduced with CytoTune - EmGFP Sendai Fluorescence Reporter at an MOI of 5 shown at 48 hours.   Green expression demonstrates the uptake of Sendai virus into the cells.
Successful iPSC reprogramming from a range of somatic cell types
The following somatic cell types have been successfully reprogrammed into iPSCs using CytoTune technology.
  • Adult and neonatal dermal fibroblasts
  • Amniotic fluid MSCs
  • Cardiac fibroblasts
  • CD34+ blood cells
  • Mammary epithelial cells
  • Nasal epithelial cells
  • Peripheral blood mononuclear cells (PBMCs)
  • Skeletal myoblasts
  • T cells
  • Umbilical vein epithelial cells
  • Urine epithelial cells
View endpoint PCR & staining
Lack of genomic integration and viral remnants allows the use of the CytoTune-iPS Sendai Reprogramming Kit in a broad range of experiments, including disease research.
Comparison of the lifecycles of non-integrating Sendai vectors and integrating vectors
Figure 3A: Schematic shows upper left corner of a cell including the cytoplasm (surrounding tan outline) and the nucleus (circular blue middle). Arrows and icons demonstrate how Sendai virus is introduced to the cell and works within it. Figure 3B: Schematic shows upper right corner of a cell including the cytoplasm (surrounding tan outline) and the nucleus (circular blue middle). Arrows and icons demonstrate how other vectors (besides Sendai virus) are introduced to the cell and work within it.
Sendai Virus (SeV) is an enveloped virus of 150–250 nm in diameter whose genome is a single chain RNA in the minus sense. Six genes coding for viral proteins are situated sequentially on the genome of the wild-type SeV in the following order.
Nucleocapsid protein (NP) forms the core nucleocapsid complex with the genome RNA. Phosphoprotein (P) is the small subunit of the RNA polymerase.  Matrix protein (M) supports the envelope structure from the inside. Fusion protein (F) fuses the viral envelope with the cell membrane when the virus enters the cell. The gene encoding the F protein is deleted from the CytoTune® -iPS Sendai Reprogramming Kit vectors, rendering them incapable of producing infectious particles from infected cells. 
Hemagglutinin-Neuraminidase (HN) recognizes the cell surface receptor, sialic acid.  Large protein (L) is the large subunit of RNA polymerase.
Because SeV infects cells by attaching itself to the sialic acid receptor present on the surface of many different cells, it can infect a wide range of cell types of various animal species. Activation of F protein by a protease is required for the virus-cell fusion process to take place. After infection, the virus goes through genome replication and protein synthesis in the cytoplasm, and then daughter virus particles are assembled and released.
Other viral vectors, such as lentivirus or retrovirus, require integration into the host genome for replication and can therefore disrupt the genome of the cells that are reprogrammed rendering iPSCs and their derivatives less safe for clinical applications or altering gene expression patterns which can compromise compound screens or disease pathway analyses. A single transduction with the CytoTune® -iPS Sendai Reprogramming Kit generates iPSCs at high efficiency with no genomic integration or viral remnants allowing the use of iPSCs and their derivatives in a broader range of research experiments.
Learn more about the CytoTune kits
Ordering Information
Catalog # Name Size Price (USD) Qty
A16517 CytoTune™-iPS 2.0 Sendai Reprogramming Kit, 1 Pack Each
3,933.65
Online Exclusive
4,225.00
Save 291.35 (7%)
A16518 CytoTune™-iPS 2.0 Sendai Reprogramming Kit, 3 Pack Each
10,054.65
Online Exclusive
10,800.00
Save 745.35 (7%)
For research use only. Not for human or animal therapeutic or diagnostic use.
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.