StemPro media
Xeno-free media specifically formulated for the growth and expansion of human mesenchymal stem cells (hMSC) and human adipose-derived stem cells (hADSC)
  • Supports hMSC and hADSC expansion for multiple passages
  • Maintains spindle-shaped morphology
  • Maintains multilineage mesoderm differentiation potential
  • Supports primary isolation of hMSCs
  • StemPro® MSC SFM XenoFree is cGMP compliant facilitating regulatory review/approval
Catalog # Name Size Price (USD) Qty
A1067501 StemPro™ MSC SFM XenoFree Each
521.90
Special offer
Online exclusive
Ends: 21-Jun-2025
614.00
Save 92.10 (15%)
Supports hMSC expansion
Expansion of hMSCs using StemPro MSC SFM XenoFree on CELLstart substrate-coated flasks is comparable to classical medium (DMEM + 10% FBS) (Figure 1).
Expansion of MSCs in StemPro MSC SFM XenoFree
Figure 1.  Expansion of MSCs in StemPro MSC SFM XenoFree.  Human Bone Marrow-derived MSCs (BM-MSCs) cultured in StemPro® MSC SFM XenoFree on CELLstart-coated flasks reveal a similar growth rate compared to DMEM + 10% MSC-Qualified FBS with long-term multi-passage capability
Better quality cells—spindle-shaped morphology
hMSCs grown in StemPro MSC SFM XenoFree medium on CELLstart substrate-coated flasks exhibit a less flattened, spindle-shaped morphology (Figure 2).
StemPro MSC SFM XenoFree medium
Figure 2. Human BM-MSC morphology.  BM-MSCs  expanded in StemPro MSC SFM XenoFree display a conserved spindle-shaped morphology after 9 passages compared to DMEM + 10% MSC-Qualified FBS. Images were captured using a 10X objective.
Maintains multilineage differentiation potential
hMSCs cultured in StemPro® MSC SFM XenoFree retain their tri-lineage mesoderm differentiation potential (Figure 3).
Multilineage differentiation with StemPro MSC medium
Figure 3.  Multilineage differentiation. Human MSCs expanded for 5 passages in StemPro® MSC SFM XenoFree retained multipotency as shown by the ability to differentiate into adipocytes, osteoblasts, and chondrocytes. Staining: adipogenesis = Oil Red O (Day 14); chondrogenesis = Alcian blue (Day 14)  osteogenesis = alkaline phosphatase (Day 21). Images were captured using a 10X objective.
Primary Isolation of hMSCs
Purified bone marrow mononuclear cells cultured in StemPro MSC SFM XenoFree supplemented with low level pooled human AB serum (2.5%) supported primary BM-MSC expansion with continual propagation (after Passage 0) under serum- and xeno-free conditions (Figure 4).
Primary Human BM-MSC Expansion
Figure 4.  Primary Human BM-MSC Expansion. Ficoll-purified BM-MNCs cultured in StemPro® MSC SFM XenoFree (on CELLstart-coated flasks) supplemented with low level pooled human AB serum (2.5%) supported primary BM-MSC expansion with continual propagation (after P0) under serum- and xeno-free conditions.
For Research Use Only. Not for use in diagnostic procedures.