The table below lists some potential problems and possible solutions that may help you troubleshoot your cell culture experiments. Note that the list below includes only the most commonly encountered problems in cell culture, and provides guidelines to solutions only. To help evaluate your results more successfully, we recommend that you consult the manuals and product information sheets provided with the products you are using as well as the published literature and books on the subject.
Obtain new stock and store in liquid nitrogen. Keep the cells in liquid nitrogen until thawing
Home made freezer stock is not viable
Freeze cells at a density recommended by the supplier
Use low-passage cells to make your own freezer stocks
Follow procedures for freezing cells exactly as recommended by the supplier. Note the freezing procedure recommended by this handbook is a general procedure provided as a guideline only.
Obtain new stock
Cells were thawed incorrectly
Follow procedures for thawing cells exactly as recommended by the supplier. Note the thawing procedure recommended by this handbook is a general procedure provided as a guideline only.
Make sure that you thaw the frozen cells quickly, but dilute them slowly using pre-warmed growth medium before plating.
Thawing medium is not correct
Use the medium recommended but the supplier. Make sure the medium is pre-warmed
Cells are too dilute
Plate thawed cells at high density as recommended by the supplier to optimize recovery.
Cells not handled gently
Freezing and thawing procedures are stressful to most cells. Do not vortex, bang the flasks to dislodge the cells (except when culturing insect cells), or centrifuge the cells at high speeds.
Glycerol used in the freezing medium was stored in light (if applicable)
If stored in light, glycerol is gets converted to acrolein, which toxic to cells. Obtain new stock.
Problem: Cells Grow Slowly
Reason
Solution
Growth medium is not correct
Use pre-warmed growth medium as recommended by the supplier.
Getting started in 3D - Setting up your lab and reviewing common pitfalls of researchers new to 3D
Part 1: Getting started in 3D: Setting up your lab and reviewing common pitfalls of 3D research Part 2: Monitoring your 3D culture (what's good and bad): How to detect problems and what a successful growth looks like
Video Player is loading.
Current Time 0:00
/
Duration 1:05:27
Loaded: 0.25%
0:00:00
Stream Type LIVE
Remaining Time -1:05:27
1x
Chapters
descriptions off, selected
captions settings, opens captions settings dialog
captions off, selected
en (Main), selected
This is a modal window.
Beginning of dialog window. Escape will cancel and close the window.
End of dialog window.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
Now Playing
Up NextGetting started in 3D - Setting up your lab and reviewing common pitfalls of researchers new to 3D
Part 1: Getting started in 3D: Setting up your lab and reviewing common pitfalls of 3D research Part 2: Monitoring your 3D culture (what's good and bad): How to detect problems and what a successful growth looks like
Now Playing
Up NextCell Culture: Cell Culture Basics
In this video, we present the basic equipment used in cell culture and proper way to set-up a laboratory. Cell culture refers to the removal of cells from an animal or plant and their subsequent growth in a favorable artificial environment.
Now Playing
Up NextAseptic Techniques: Cell Culture Basics
The handbook and videos provide an introduction to cell culture, with a focus on aseptic technique, microbial contamination, laboratory safety and cell culture experiments.
Now Playing
Up NextFreezing Cells: Cell Culture Basics
This cell culture workflow video focuses on steps pertaining to freezing cells. It also contains useful information, troubleshooting guides and protocols for the experienced researcher as well.
Now Playing
Up NextPassaging Cells: Cell Culture Basics
This video provides an introduction to cell culture, with a focus on maintaining cell health throughout the processes of culturing, freezing, thawing and passaging cells. In this video, we focus on how to passage cells.
Now Playing
Up NextThawing Cells: Cell Culture Basics
This video provides an introduction to cell culture, with a focus on maintaining cell health throughout the processes of culturing, freezing, thawing and passaging cells. In this video, we focus on how to thaw cells.