Stunningly easy western blot imaging
(See a list of the products featured in this article.)
Despite the rapid pace of technological advances in life science research, nucleic acid gels, protein gels, and western blots continue to be fundamental laboratory tools with the ability to move a project forward or push it in a whole new direction. The Invitrogen iBright Imaging Systems are high-performance instruments for capturing images and analyzing data from gels and blots. Designed with a streamlined intuitive interface and advanced automated features, the iBright systems are easy to use for researchers at all experience levels.
iBright CL1000 and iBright FL1000 Imaging Systems
Two iBright instruments are available: the iBright CL1000 system, for imaging and documenting chemiluminescent western blots and colorimetrically stained protein and nucleic acid gels, and the iBright FL1000 system (Figure 1), which features the same imaging modes as the iBright CL1000 system but also offers fluorescent gel and blot imaging capability using visible and near-IR excitation and emission channels (Table 1).
Advantages of the iBright Imaging Systems
  • Powerful 9.1 megapixel cooled 16-bit CCD camera—for capturing crystal-clear images with robust imaging potential (Figure 2).
  • Push-button optimized exposure with Invitrogen Smart Exposure technology—for rapid determination of optimal exposure times, minimizing the need to repeat exposures to get the desired signal.
  • Advanced automated features—allowing hassle-free sample alignment, focus, and zoom, as well as automatic on-board data analysis for instantaneous lane and band identification and molecular weight marker overlay.
  • Long-life green LED-based transilluminator—providing a light source that can effectively excite popular DNA dyes (e.g., ethidium bromide and Invitrogen SYBR Green dyes), without exposing users to harmful UV radiation.
  • Cloud connectivity with Invitrogen iBright Analysis Software— enabling export and storage of data, as well as the ability to access, review, analyze, and share data through the web-based Thermo Fisher Cloud platform.
  • Extensive service and support—from the on-site Invitrogen SmartStart orientation (included with every system) to our highly responsive technical and field support teams and comprehensive post-warranty service plans.
The iBright FL1000 Imaging System
Figure 1. The iBright FL1000 Imaging System.
Table 1. Imaging modes of the iBright FL1000 and iBright CL1000 systems.
Imaging capabilityWhat kind of signal can be captured?
Protein gelColorimetric staining of gels (e.g., Coomassie and silver stains) and membranes (e.g., Ponceau S and MemCode stains)
Nucleic acid gelEthidium bromide and SYBR dye staining
Chemiluminescent blotChemiluminescence detection using all popular horseradish peroxidase (HRP) and alkaline phosphatase (AP) substrates (e.g., SuperSignal and WesternBreeze substrates)
Fluorescent blot *Fluorescence detection using RGB (visible range) and near-IR fluorophores (e.g., Alexa Fluor and Alexa Fluor Plus dyes and DyLight dyes)
* iBright FL1000 Imaging System only.
 iBright Imaging Systems feature a powerful 9.1 megapixel camera
Figure 2. iBright Imaging Systems feature a powerful 9.1 megapixel camera for sensitive signal detection. Two-fold serial dilutions of HeLa cell lysate (starting at 80 μg/lane) were loaded and run on Invitrogen Novex Tris-glycine gels, transferred, and probed with antibodies against DDX3 or Ku80 protein. Blots were then probed with corresponding HRP-conjugated secondary antibodies, developed with Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate, and visualized (using 10 sec exposures) on either the Invitrogen iBright FL1000 Imaging System or another imaging device with a lower-quality, 4.1 megapixel camera.
iBright FL1000 system for fluorescence applications
The iBright FL1000 Imaging System employs the same green LEDbased transilluminator as the iBright CL1000 Imaging System, as well as two high-quality long-life epi-LEDs for fluorescence imaging applications. The broad-spectrum white epi-LED can be used to excite most green-, red-, and far-red–fluorescent dyes, including antibody conjugates labeled with Invitrogen Alexa Fluor and Alexa Fluor Plus dyes, Thermo Scientific DyLight dyes, and Invitrogen WesternDot dyes. The second epi-LED emits longer-wavelength light and is optimized for near-IR fluorophores such as Alexa Fluor 790 and Alexa Fluor Plus 800 dyes, DyLight 800 dye, and WesternDot 800 dye.
Light from these LED sources, in conjunction with both excitation and emission filters, enables many possible reagent options for nucleic acid gel, protein gel, and western blot imaging applications. The iBright FL1000 model features five fluorescence channels, three in the visible fluorescence range and two in the near-IR fluorescence range (Table 2). Up to four channels can be multiplexed simultaneously when imaging a single western blot or set of four mini blots (Figure 3). Multiplex western blotting allows the study of multiple proteins in a single blot, while also providing more accurate comparisons of experimental data and controls. Smart Exposure technology further improves acquisition of multiplex fluorescence western blot data because its advanced auto-exposure functionality can help to optimize signal-to-noise ratios for each fluorescence channel separately.
Table 2. Filter sets on the iBright FL100 Imaging System.
ExcitationFilter (nm)EmissionFilter (nm)Example of a compatible Alexa Fluor Plus dye
EX1455–485EM1510–555Alexa Fluor Plus 488
EX2515–545EM2565–615Alexa Fluor Plus 555
EX3610–635EM3675–720Alexa Fluor Plus 647
EX4655–680EM4725–750Alexa Fluor Plus 680
EX5745–765EM5810–850Alexa Fluor Plus 800
Multiplexed fluorescence western blots visualized on the iBright FL1000 Imaging System
Figure 3. Multiplexed fluorescence western blots visualized using the iBright FL1000 Imaging System. (A) 3-color blot. (B) 4-color high-throughput image with 4 mini blots. (C) 2-color high-throughput image with 4 mini blots.
See the iBright Imaging Systems for yourself
To learn more about the iBright FL1000 and iBright CL1000 Imaging Systems, download the iBright Imaging Systems brochure, which shows a variety of data acquired and analyzed using the iBright systems, a list of dyes and stains compatible with the iBright imaging modes, and complete instrument specifications. Find out how these high-performance instruments can enhance gel and western blot analysis in your laboratory.
Catalog # Name Size Price (USD) Qty
A32749 iBright™ CL1000 Imaging System 1 system
44,120.00
LC5615 iBright™ Prestained Protein Ladder, 2 x 250 μL Each
328.65
Online Exclusive
343.00
Save 14.35 (4%)
Article download
Download a printer-friendly version of this article.
Download now
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
For Research Use Only. Not for use in diagnostic procedures.