Assess cell health and function in 2D and 3D cell cultures
(See a list of the products featured in this article.)
3D cell models are now increasingly being adopted for cancer, immuno-oncology, neuroscience, and other research areas because their microenvironments more closely resemble the microanatomy of in vivo systems (organized tissues, organs, and tumors), which contain a complex and dynamic set of cell types, chemical gradients, and extracellular matrix (ECM) components. The complexity of 3D cell structures can present a challenge when using cell health assays and protocols originally developed for monolayer (2D) cell cultures. Here we highlight a few basic considerations for adapting and optimizing our microplate-based 2D cell culture viability assays for spheroid cultures, focusing on the optimization of reagent concentration and incubation time.
Determine the optimal reagent concentration
The optimal reagent concentration for spheroid cultures will provide a high assay-specific signal with the lowest possible nonspecific background, resulting in a high signal-to-noise (S/N) ratio. An assay with a high S/N ratio shows greater sensitivity for detecting changes in cell health due to cell treatments, such as exposure to drugs or test compounds. To demonstrate this effect, we assayed spheroids derived from human lung epithelial cells (A549 cells) using the Invitrogen CyQUANT XTT Cell Viability Assay, a colorimetric microplate assay originally developed to assess 2D cell viability as a function of redox potential (Figure 1). We compared the recommended reagent concentration (1X) with a 2X concentration, in the absence or presence of increasing concentrations of the cytotoxic drug gambogic acid, and found that doubling the reagent concentration increased the S/N ratio, and thus the assay sensitivity. We recommend testing a wide range of reagent concentrations to find the most appropriate one for your 3D assay.
line graph of absorbance versus gambogic acid concentration for spheroids assayed using 1X or 2X XTT
Figure 1. Determine the optimal reagent concentration. Human lung epithelial cells (A549 cells) were plated in Thermo Scientific Nunclon Sphera 96U-Well Microplates at 5,000 cells/well in complete MEM for 19 hr to allow spheroid formation. The surface of these plates exhibits extremely low ECM binding properties, which inhibits cell attachment and promotes spheroid formation. The A549 cell spheroids were then treated with 7 concentrations of gambogic acid for 26 hr and assayed for cell health with the Invitrogen CyQUANT XTT Cell Viability Assay using the recommended (1X) or 2X reagent concentration. All measurements were made using the Thermo Scientific Varioskan LUX Multimode Microplate Reader.
Determine the optimal reagent incubation time
The optimal reagent incubation time for spheroid cultures can be determined by running a time-course experiment and plotting assay signal as a function of incubation time. Figure 2 shows an example of such a time-course experiment using an established microplate viability assay that determines redox potential by measuring the reduction of resazurin to the highly fluorescent (and intensely colored) resorufin. Using the Invitrogen PrestoBlue HS Cell Viability Reagent with various incubation times, we compared the fluorescence generated by A549 cells in a 2D monolayer with that of A549 cell–derived spheroids. The assay protocol recommends incubation times for 2D cell culture of 10 minutes to 3 hours, within the linear range of the time-course curve. Based on our experiments with spheroids, we recommend extending the incubation time—to between 5 and 10 hours—to maximize signal and stay within the linear range of this curve.
line graph of fluorescence versus incubation time for monolayers or spheroids assayed with PrestoBlue viability reagent
Figure 2. Determine the optimal reagent incubation time. A549 cell spheroids (prepared as described in Figure 1) and A549 cell monolayers were assayed at various incubation times with Invitrogen PrestoBlue HS Cell Viability Reagent. All measurements were made using the Thermo Scientific Varioskan LUX Multimode Microplate Reader.
Catalog # Name Size Price (USD) Qty
X12223 CyQUANT™ XTT Cell Viability Assay, 450 nm and 660 nm 1 kit
403.65
Online Exclusive
416.00
Save 12.35 (3%)
174925 96 Well Round (U) Bottom Plate, Sphera Low-Attachment Surface, Pack of 1 Case of 8
206.65
Online Exclusive
224.00
Save 17.35 (8%)
P50200 PrestoBlue™ HS Cell Viability Reagent, 25 mL Each
350.65
Online Exclusive
361.00
Save 10.35 (3%)
P50201 PrestoBlue™ HS Cell Viability Reagent, 100 mL Each
715.65
Online Exclusive
738.00
Save 22.35 (3%)
VLBLATD0 Varioskan™ LUX Multimode Microplate Reader, luminescence, time-resolved fluorescence, AlphaScreen, top and bottom, no dispenser Each -
For Research Use Only. Not for use in diagnostic procedures.