Complete growth medium
ComponentCat. No.
Gibco DMEM, with GlutaMAX SupplementA3635201
10% Gibco FBSA3160401
Proper culture techniques and procedures are an essential part of ensuring successful transfection. Subculturing, also referred to as passaging, is the removal of medium and transfer of cells from a culture into fresh growth medium, in order to propagate the cells. 
See our product list for this protocol
Transfecting plasmid DNA into HepG2 cells
Brightfield image of HepG2 cells in culture, prior to transfection.
Tip: HepG2 cells tend to grow in clusters. It is critical to ensure dissociation has occurred to yield a single-cell suspension when passaging and plating the cells. Having a single-cell suspension is also very important to ensure efficient transfection.
Passaging
  • Maintain cells in T-75 flasks.
  • Use Gibco TrypLE dissociation reagent.
  • Passage cells every 3–4 days to ensure that they do not enter senescence.
  • Transfection of cells should be performed only between passages 5 and 25 post-thaw.
  • If designing an experiment that involves transfection, ensure that setup coincides with a cell passage.
  • Plate cells for transfection only 1 day before the experiment.
Tip: Trypsinize HepG2 cells with TrypLE dissociation reagent for 10–15 min in a 37°C incubator. Then add 10 mL of growth medium and use a 10 mL serological pipette with a 200 μL tip on the end to pipette the cell suspension up and down at least 5 times. This step is the most critical to ensure single cells for accurate counting and plating.
Seeding cells for transfection
  • The day before transfection, dissociate cells that are 80–90% confluent in a T-75 flask.
  • Count the cells using standard trypan blue exclusion.
    • ––Important: The cell number and concentration determined can vary significantly depending on what method is used for counting; it is important to be consistent and use a single method throughout an experiment.
  • The cell culture must have >90% viability and be 75–80% confluent on the day of transfection.
    • ––Important: If cells are not at the right confluence, do not wait until the next day to perform transfection, as this can significantly affect transfection efficiency.
  • Seed 1.1 x 105 cells in 500 μL growth medium for a single well of a 24-well plate.
Transfection protocol
ComponentCat. No.
Invitrogen Lipofectamine 3000 Transfection ReagentL3000008
Gibco Opti-MEM I Reduced Serum Medium31985062
Thermo Scientific Nunc Multidish, 24-Wells, Cell Culture Treated142475
On the day of transfection, which should be 1 day following cell plating, perform the following steps, which have been optimized for a single well of a 24-well plate using Invitrogen Lipofectamine 3000 Transfection Reagent:
StepTubeComplexation componentsAmount per well
(24-well)
1Tube 1Opti-MEM I medium25 μL
Lipofectamine 3000 reagent0.75 μL
2Tube 2Opti-MEM I medium25 μL
DNA amount (DNA concentration should be 0.5–5 μg/μL)500 ng
P3000™ reagent1 μL
3Add tube 2 solution to tube 1 and mix well
4Incubate mixture from step 3 at room temperature for 10–15 min
5Add 50 μL of complex from step 4 to cells;
gently swirl plate to ensure homogeneous distribution of complex to the entire well
Transfection efficiency analysis
At 48 hr following transfection of a GFP reporter construct, cells were evaluated via microscopy and flow cytometry. To assess transfection efficiency, cells were first visualized via fluorescence microscopy for qualitative assessment of protein expression, morphology, and viability (Figure 1). Cells were then prepared for flow cytometry by aspirating the medium and replacing it with 250 μL of a 7:3 mixture of TrypLE reagent:1X DPBS. Cells were incubated at 37°C for 10 min and then pipetted up and down to ensure single cells for flow cytometry analysis.
Posttransfection analysis of fluorescence cells
Posttransfection analysis of bright-fields cells
Figure 1. Posttransfection analysis of cells. (A) Fluorescence and (B) bright-field images demonstrating 65–75% transfection efficiency.
Tips and tricks
  • Decreasing the serum content of the culture medium (to <10%) at the time of transfection is acceptable, but replace with complete growth medium within 4–24 hr post-transfection.
  • Antibiotics can be used during transfection.
  • Prior to flow cytometry, visualize cells under a bright-field microscope to verify dissociation following incubation with TrypLE reagent.
Scaling up or down Lipofectamine 3000 reagent transfections
Use the following table to scale the volumes for your transfection experiment. The most common sizes are listed below.
Culture vesselMultiplication factor*Shared reagentsDNA transfectionsiRNA transfection
Growth mediumOpti-MEM medium for complexingDNAP3000 reagentLipofectamine 3000 reagent**siRNALipofectamine 3000 reagent**
96-well0.2100 μL2 x 5 μL100 ng0.1 µL0.15 µL3 pmol0.3 µL
48-well0.5250 μL2 x 12.5 μL0.25 µg0.5 µL0.375 µL7.5 pmol0.75 µL
24-well1500 μL2 x 25 μL0.5 µg1 µL0.75 µL15 pmol1.5 µL
12-well21 mL2 x 50 μL1 µg2 µL1.5 µL30 pmol3 µL
6-well52 mL2 x 125 μL2.5 µg5 µL3.75 µL75 pmol7.5 µL
60 mm11.055 mL2 x 250 μL5.5–11 µg11–22 µL8.5 µL166 pmol17 µL
10 cm28.9510 mL2 x 500 μL14–28 µg28–56 µL22 µL434 pmol43 µL
T-7539.4715 mL2 x 750 μL20–40 µg40–80 µL30 µL592 pmol59 µL
T-17592.1135 mL2 x 1.75 mL46–96 µg92–180 µL69 µL1,382 pmol138 µL
* After determining the optimum reagent amount, use the multiplication factor to determine the reagent amount needed for your new plate format.
** Optimum amount needed is determined from the protocol for Lipofectamine 3000 Transfection Reagent.
Ordering information
Gibco Liver Cancer Starter Kit
For your convenience, the essential components of this protocol are now available in the Gibco Liver Cancer Starter Kit. The kit includes: basal medium, FBS, Lipofectamine 3000 reagent, Opti-MEM medium, and TrypLE reagent. The kit is available at thermofisher.com/cancercellculture. For additional components required for the protocol see ordering table below.
Catalog # Name Size Price (USD) Qty
142475 24 Well Plate, TC Surface, Pack of 1 Case of 75
400.65
Online Exclusive
456.00
Save 55.35 (12%)
For Research Use Only. Not for use in diagnostic procedures.