Human Mesenchymal Stem Cells Lipofectamine® 2000
Introduction
Lipofectamine® 2000 Reagent is a proprietary formulation that facilitates highly efficient delivery of Stealth™ RNAi or short interfering RNA (siRNA) to mammalian cells for RNAi analysis. This reference provides general guidelines and an optimized procedure to transfect Stealth™ RNAi (or siRNA) into human MSC mesenchymal stem cells (Cambrex BioScience, Cat. No. PT-2501) using Lipofectamine® 2000 (Cat. No. 11668-027).
Note: While transfection conditions have been optimized to allow highly efficient delivery of Stealth™ RNAi into MSC cells, other factors related to the target gene of interest including the transcription rate of the target gene, the stability of the resulting protein, and efficacy of the Stealth™ RNAi sequence chosen can influence the degree of target gene knockdown observed. Take these factors into consideration when designing your RNAi experiment.
Guidelines for Transfection
Follow these important guidelines when transfecting Stealth™ RNA (or siRNA) into MSC cells using Lipofectamine® 2000:
  1. Use 400 nM Stealth™ RNAi (or siRNA) complexed with 2 µg/ml Lipofectamine® 2000 (stock solution is 1 mg/ml) for transfection. To increase accuracy and reduce assay variability, we recommend performing transfection in triplicate for each sample condition.
  2. Transfect MSC cells at 70% confluence.
  3. Do not add antibiotics to the medium during transfection as this reduces transfection efficiency and causes cell death.
  4. Use Opti-MEM® I Reduced Serum Medium (Cat. No. 31985-062) to dilute Lipofectamine® 2000 and Stealth™ RNAi (or siRNA) prior to complex formation.
Materials
Have the following reagents on hand before beginning:
    * MSC cells maintained in Mesenchymal Stem Cell Basal Medium (MSCBM; Cambrex BioScience, Cat. No. PT-3238) supplemented with MSCGM SingleQuots (Cambrex BioScience, Cat. No. PT-4105)
Note: Use low-passage cells (passage 5 or lower); make sure that cells are healthy and greater than 90% viable before transfection.
    * Stealth™ RNAi (or siRNA) of interest (20 µM in annealing buffer)
    * Lipofectamine® 2000 Reagent (store at +4°C until use)
    * Opti-MEM® I Reduced Serum Medium (pre-warm to 37°C before use)
    * Appropriate tissue culture plates and supplies
Ordering Information
Catalog # Name Size Price (USD) Qty
11668027 Lipofectamine™ 2000 Transfection Reagent, 0.75 mL Each
545.65
Online Exclusive
590.00
Save 44.35 (8%)
11668019 Lipofectamine™ 2000 Transfection Reagent, 1.5 mL Each
842.65
Online Exclusive
924.00
Save 81.35 (9%)
Protocol
Use this procedure to transfect Stealth™ RNAi (or siRNA) into MSC cells using Lipofectamine® 2000 in a 24-well format. For other formats, see the table in Recommended Reagent Amounts and Volumes for the appropriate reagent amounts to add. Tip: To reduce well-to-well variability when transfecting multiple replicates (e.g. triplicates), proportionally scale up the reagent volumes to form complexes (Step 2), then aliquot an equal volume of complexes into each well.
  1. One day before transfection, plate 3 x 104 MSC cells in 400 µl of growth medium without antibiotics per well. Cells should be 70% confluent at the time of transfection.
  2. For each transfection sample, prepare Stealth™ RNAi-Lipofectamine® 2000 complexes as follows:
    • Dilute 200 pmol of Stealth™ RNAi (i.e. 10 µl of 20 µM Stealth™ RNAi) in 50 µl of Opti-MEM® I   Reduced Serum Medium. Mix gently.
    • Mix Lipofectamine™ 2000 gently before use, then dilute 1 µl in 50 µl of Opti-MEM® I Reduced Serum Medium. Mix gently and incubate for 15 minutes at room temperature.    
    • After the 15-minute incubation, combine the diluted Stealth™ RNAi and the diluted Lipofectamine® 2000 (total volume = 111 µl). Mix gently and incubate for 15 minutes at room temperature to allow complexes to form (solution may appear cloudy).
  3. Add the 111 µl of Stealth™ RNAi-Lipofectamine® 2000 complexes to each well containing cells and medium. Mix gently by rocking the plate back and forth.
  4. Incubate the cells at 37°C in a humidified CO2  incubator for 16-24 hours as appropriate until you are ready to assay for gene knockdown.
Recommended Reagent Amounts and Volumes
To transfect MSC cells in different tissue culture formats, vary the amounts of Stealth™ RNAi (or siRNA), Lipofectamine® 2000, cells, and medium used in proportion to the relative surface area, as shown in the table. Note: 20 µM Stealth™ RNAi or siRNA = 20 pmol/µl.
Culture vessel
Relative surface area (vs. 24-well)
Cells plated per well
Volume of plating medium
Stealth RNAi (pmol) in media volume (µl)
Lipofectamine® 2000 (µl) in media volume (µl)
48-well
          0.4
   1.5 x 10 4
       200 µl
       80 pmol in 25 µl
     0.4 µl in 25 µl
24-well
            1
      3 x 10 4
       400 µl
     200 pmol in 50 µl
        1 µl in 50 µl
  6-well
            5
   1.5 x 10 5
          2 ml
 1000 pmol in 250 µl
      5 µl in 250 µl
LT077