Background
The pharmaceutical and biotechnology industry’s goal is to discover therapeutic agents that are both safe and effective at treating or preventing diseases. Compounds identified as selective and potent in the early drug discovery phase are progressed to preclinical drug development for further evaluation. It is estimated that over 10% of drugs fail in clinical trials due to pharmacokinetic reasons (1), and the US Food and Drug Administration (FDA) guidelines emphasize the identification of metabolic pathways, relevant metabolites and potential drug-drug interactions for new chemical entities (NCEs) where metabolism is the principal route of elimination (2) Consequently, drug metabolism studies are a critical constituent of any drug development program.
The primary site of metabolism for many drugs is the liver. Liver-derived systems such as liver slices, sub-cellular liver fractions, and intact hepatocytes are typically utilized to assess the metabolism of NCEs. Intact hepatocytes contain the cytochrome P450’s (CYPs), other non-P450 enzymes, and phase II enzymes such as sulfo- and glucuronosyltransferases, and thus represent a prime model system for studying drug disposition in vitro (3). NCEs can be screened and rank-ordered according to metabolic half-life estimates or in vitro intrinsic clearance values (Cl
int,
in vitro) obtained from metabolic stability studies. Moreover, metabolic screening assays enable drug developers to focus on the improvement of compounds through structural activity relationships (SAR) and prevent the progression of labile compounds to more costly in vivo studies. Given that cryopreserved hepatocytes retain enzymatic activities similar to those of fresh hepatocytes and offer convenience to the end user (4), the utility of cryopreserved hepatocytes in efforts to define a drug’s disposition in vitro is advantageous as compared to other model systems.
Important notes
- Review this protocol to ensure y ou have all the necessary reagents and equipment prior to starting the procedure. Once thawed, cryopreserved hepatocytes must be used immediately and will not maintain viability if refrozen.
- Use universal safety precautions and appropriate biosafety cabinet when handing primary hepatocytes.
Critical materials and reagents
- Cryopreserved hepatocytes for suspension use, such as Life Technologies Cat. No. HMCS1S or HMCS2S (check website for additional catalog numbers), enough for 6 x 106 cells per plate
- Williams’ Medium E, 500 mL, Life Technologies Cat. No. CM6000
- Hepatocyte Maintenance Supplement Pack (Serum-free), 1 kit, Life Technologies Cat. No. CM4000
- 12-well non-coated plates (Greiner Bio-One, Cat. No. 665 180 or equivalent).
- 15-mL conical tubes (1 per compound)
- Compound stocks: test articles (TA) and positive controls (PC). Suitable positive controls may include:
- midazolam
- phenacetin
- testosterone
- dextromethorphan
- (S)-mephenytoin
- 7-hydroxycoumarin
- Stop solution
Equipment
- 37°C water bath
- 37°C / 5% CO2 humidified incubator
- Orbital shaker placed inside incubator
For questions related to this protocol, contact us at:
Email: hepaticproducts@lifetech.com
Phone: +1 919 237 4500 (Toll)
Phone: +1 866 952 3559 (U.S. Toll-free)