Protocols
Introduction
Intended use
Dynabeads M-280 Streptavidin are ideal for numerous applications, including purification of proteins and nucleic acids, protein interaction studies, immunoprecipitation, immunoassays, phage display, biopanning, drug screening, and cell isolation.
Principle
Add Dynabeads to a sample containing biotinylated molecules e.g., peptides, proteins, antibodies, sugars, lectins,  oligonucleotides, DNA/RNA. During a short incubation, the biotinylated molecule will bind to the beads. Separate the molecule-bead complex with a Dynal magnet. Capture, washing and detection can be optimized for manual or automated use. With indirect capture, mix the biotinylated molecule with the sample to capture the molecule-target complex before adding Dynabeads. Indirect target capture is an advantage when molecule-target kinetics are slow, affinity is weak, molecule concentration is low or molecule-target binding requires optimal molecule orientation and true liquid-phase kinetics.
Description of materials
Dynabeads Streptavidin are uniform, superparamagnetic beads of 2.8 μm in diameter with a streptavidin monolayer covalently coupled to the surface. This layer ensures negligible streptavidin leakage while the lack of excess adsorbed streptavidin ensures batch consistency and reproducibility of results.
Materials supplied
Dynabeads M-280 Streptavidin are supplied as a suspension containing 10 mg (6–7 x 108)
Dynabeads per mL, dissolved in phosphate buffered saline (PBS) pH 7.4, containing 0.1%
BSA and 0.02% NaN3 as preservatives. Available in three volumes:
  • 2 mL (Cat. No. 112.05D)
  • 10 mL (Cat. No. 112.06D)
  • 100 mL (Cat. No. 602.10)
Additional materials required
Magnet for manual protocols.
Table 1. Recommended buffers and solutions.
For coupling of nucleic acidsFor Dynabeads treatment before RNA manipulationsFor coupling of protein or other molecules
Binding and washing (B&W) Buffer (2x):
  • 10 mM Tris-HCl (pH 7.5)
  • 1 mM EDTA
  • 2 M NaCl
Solution A:
  • DEPC-treated 0.1 M NaOH
  • DEPC-treated 0.05 M NaCl
Solution B:
  • DEPC-treated 0.1 M NaCl
PBS buffer pH 7.4 These buffers can also be used for your application if needed:
  • PBS/BSA (PBS, pH 7.4 containing 0.1% (w/v) BSA)
  • PBST (PBS, pH 7.4 containing 0.01% (v/v) Tween™ 20
The salt concentration and pH (typically 5–9) of the chosen binding/washing buffers can be varied depending on the type of molecule to be immobilized. Beads with immobilized molecules are stable in common buffers.
Mixing device with tilting and rotation.
Materials
Additional material needed
  • Magnetic device (Dynal MPC, Magnetic Particle Concentrator).
  • Mixing/rotation device
  • Vortex mixer
  • Test tubes and pipettes
  • Buffers/solutions
  • Primary antibody
Recommended buffers/solutions
All reagents used should be analytical grade. Isotonic (PBS) pH 7.2–7.6 We recommend the following PBS (pH 7.4):
  • NaH2PO4 x H2O 0.16 g
  • Na2HPO4 x 2 H2O 0.98 g
  • NaCl 8.10 g
  • Distilled water to 1 liter
PBS/BSA: Add 0.1% BSA fraction V (final conc.) to PBS. 0.02% NaN3 may be added as a preservative for storage, if needed.
Technical information
Binding capacity
Both the size of the molecule to be immobilized and the biotinylation procedure will affect the binding capacity. Large as well as small biotinylated molecules can be immobilized. The capacity for biotinylated molecules depends on steric availability and charge interaction between bead and molecule and between molecules. There are two or three biotin binding sites available for each streptavidin molecule on the surface of the bead after immobilization.
  • Optimize the quantity of beads used for each individual application by titration.
  • Use up to two-fold excess of the binding capacity of the biotinylated molecule to saturate  streptavidin.
  • Binding efficiency can be determined by comparing molecule concentration before and after coupling.
Ordering Information
Catalog # Name Size Price (USD) Qty
11205D Dynabeads™ M-280 Streptavidin, 2 mL Each
615.40
Special offer
Online exclusive
Ends: 01-Jun-2025
724.00
Save 108.60 (15%)
11206D Dynabeads™ M-280 Streptavidin, 10 mL Each
1,959.25
Special offer
Online exclusive
Ends: 01-Jun-2025
2,305.00
Save 345.75 (15%)
60210 Dynabeads™ M-280 Streptavidin, 100 mL Each
11,457.00
Special offer
Online exclusive
Ends: 01-Jun-2025
12,730.00
Save 1,273.00 (10%)
General Guidelines
  • Keep the tube on the magnet for 2 min to ensure that all the beads are collected on the tube wall.
  • For diluted samples, increase the incubation time or isolate in smaller batches using the same beads in each batch.
  • Avoid air bubbles during pipetting.
  • Free biotin in the sample will reduce the binding capacity of the beads.
  • A disposable separation column or a spin column will remove unincorporated biotin.
  • For some applications it can be an advantage to add a detergent such as 0.01– 0.1% Tween™ 20 to the washing/binding buffers to reduce non-specific binding.
  • Run the PCR with limiting concentrations of biotinylated primer, or remove free biotinylated primer by ultrafiltration, microdialysis or other clean-up protocols.
  • Optimize the quantity of beads used for each individual application by titration.
  • Use up to two-fold excess of the binding capacity of the biotinylated molecule to saturate streptavidin.
  • Binding efficiency can be determined by comparing molecule concentration before and after coupling.
  • lifetechnologies.com.
Protocols
Recommended Washing Buffers
  • Nucleic acid applications: 1X B&W Buffer (see Table 1 for recipe). Dilute to 1X B&W Buffer with distilled water.
  • Antibody/protein applications: PBS, pH 7.4
Wash Dynabeads
Calculate the amount of beads required based on their binding capacity (see Table 2), and transfer the beads to a new tube.
1. Resuspend the Dynabeads in the vial (i.e. vortex for >30 sec, or tilt and rotate for 5 min).
2. Transfer the desired volume of Dynabeads to a tube.
3. Add an equal volume of Washing buffer, or at least 1 mL, and mix (vortex for 5 sec, or keep on a roller for at least 5 min).
4. Place the tube on a magnet for 1 min and discard the supernatant.
5. Remove the tube from the magnet and resuspend the washed Dynabeads in the same volume of washing buffer as the initial volume of Dynabeads taken from the vial (step 2).
Table 2 - Typical binding capacities for one mg of Dynabeads.
Biotinylated targetBinding/mL
Free Biotin [pmol]650 – 900
Biotinylated peptides [pmol]~ 200
Biotinylated antibody [μg]up to 10
ds DNA [μg] *~ 10
ss oligonucleotides [pmol] *~ 200
* Oligonucleotides and DNA fragments
For oligonucleotides, capacity is inversely related to molecule size (number of bases). Reduced binding capacity for large DNA fragments may be due to steric hindrance.
Dynabeads for RNA Manipulation
As Dynabeads Streptavidin are not supplied in RNase-free solutions, perform the following steps after washing for RNA applications:
1. Wash the beads twice in Solution A for 2 min. Use the same volume of Solution A
as the initial volume of Dynabeads taken from the vial or larger.
2. Wash the beads once in Solution B. Use the same volume of beads as in step 5.
3. Resuspend the beads in Solution B.
The beads are now ready to be coated with the biotinylated molecule of your choice.
Immobilization Protocol
Wash the Dynabeads according to “Wash Dynabeads” section before use.
1. Add the biotinylated molecule to the washed Dynabeads.
2. Incubate for 15–30 min at room temperature with gentle rotation of the tube.
3. Place the tube in a magnet for 2–3 min and discard the supernatant.
4. Wash the coated beads 3–4 times in washing buffer.
5. Resuspend to desired concentration in a suitable buffer for your downstream use.
Here are some examples of immobilization protocols for specific applications.
Immobilize Nucleic Acids
1. Resuspend beads in 2X B&W Buffer to a final concentration of 5 μg/μL (twice original volume).
2. To immobilize, add an equal volume of the biotinylated DNA/RNA in distilled water to dilute the NaCl concentration in the 2 B&W Buffer from 2 M to 1 M for optimal binding.
3. Incubate for 15 min at room temperature using gentle rotation. Incubation time depends on the nucleic acid length: short oligonucleotides (<30 bases) require max. 10 min. DNA fragments up to 1 kb require 15 min.
4. Separate the biotinylated DNA/RNA coated beads with a magnet for 2–3 min.
5. Wash 2–3 times with a 1X B&W Buffer.
6. Resuspend to the desired concentration. Binding is now complete. Resuspend the beads with the immobilized DNA/RNA fragment in a buffer with low salt concentration, suitable for downstream applications.
Immobilize Antibodies/Proteins
1. Incubate the beads and biotinylated antibodies in PBS for 30 min at room temperature using gentle rotation.
2. Separate the antibody-coated beads with a magnet for 2–3 min.
3. Wash the coated beads 4–5 times in PBS containing 0.1% BSA.
4. Resuspend to the desired concentration for your application.
Release Immobilized Biotinylated Molecules
The biotin-streptavidin bond is broken by harsh conditions. 5 min incubation at 65°C or 2 min at 90°C in 10 mM EDTA pH 8.2 with 95% formamide will typically dissociate >96% of immobilized biotinylated DNA. Alternatively, boil the sample for 5 min in 0.1% SDS for protein dissociation. Please note that proteins will be denatured by such treatment and Dynabeads Streptavidin can not be re-used. It has also been reported that the biotin-streptavidin interaction can be broken by a short incubation in nonionic water at a temperature above 70°C.
Automation
Magnetic separation and handling using Dynabeads can easily be automated using our KingFisher Separation Systems.
Reference
  1. Holmberg et al. (2005) The biotin-Streptavidin interaction can be reversibly broken using water at elevated temperatures. Electrophoresis 26, 501-510.
112.05D06D.indd   Rev 014    May-5-2007