Search Thermo Fisher Scientific
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
L-Arginine hydrochloride | 211.0 | 126.0 | 0.5971564 |
L-Cystine 2HCl | 313.0 | 31.28 | 0.099936105 |
L-Histidine hydrochloride-H2O | 210.0 | 42.0 | 0.2 |
L-Isoleucine | 131.0 | 52.0 | 0.39694658 |
L-Leucine | 131.0 | 52.0 | 0.39694658 |
L-Lysine hydrochloride | 183.0 | 72.5 | 0.39617488 |
L-Methionine | 149.0 | 15.0 | 0.10067114 |
L-Phenylalanine | 165.0 | 32.0 | 0.19393939 |
L-Threonine | 119.0 | 48.0 | 0.40336135 |
L-Tryptophan | 204.0 | 10.0 | 0.04901961 |
L-Tyrosine | 181.0 | 36.0 | 0.19889502 |
L-Valine | 117.0 | 46.0 | 0.3931624 |
Vitamins | |||
Choline chloride | 140.0 | 1.0 | 0.007142857 |
D-Calcium pantothenate | 477.0 | 1.0 | 0.002096436 |
Folic Acid | 441.0 | 1.0 | 0.0022675737 |
Niacinamide | 122.0 | 1.0 | 0.008196721 |
Pyridoxal hydrochloride | 204.0 | 1.0 | 0.004901961 |
Riboflavin | 376.0 | 0.1 | 2.6595744E-4 |
Thiamine hydrochloride | 337.0 | 1.0 | 0.002967359 |
i-Inositol | 180.0 | 2.0 | 0.011111111 |
Inorganic Salts | |||
Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 200.0 | 1.8018018 |
Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 97.67 | 0.8139166 |
Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
Sodium Chloride (NaCl) | 58.0 | 6800.0 | 117.24138 |
Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 140.0 | 1.0144928 |
Other Components | |||
D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
Phenol Red | 376.4 | 6.0 | 0.01594049 |
Sodium Succinate | 270.0 | 100.0 | 0.37037036 |
Succinic acid | 118.0 | 75.0 | 0.63559324 |
1. Eagle, H. (1959) Science 130:432.
2. Lolait, S.J., Harmer, J.H., Dahl, D., et al., (1983) Aust. J. Exp. Biol. Med. Sci., 61:439. (rat astrocytes).
3. Norris, S.J. and Edmondson, D.G. (1986) Sex. Transm. Dis., 13:207 (Rabbit epithelial cells).