Search Thermo Fisher Scientific
Low Serum Growth Supplement (LSGS) is a sterile, concentrated (50X) solution intended for use as one component in a complete culture environment for the growth of human dermal fibroblasts and endothelial cells derived from large vessels. The concentration of FBS is 95%.
Catalog Number(s)
S00310Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Proteins | |||
Basic FGF | 0.15 | Infinity | |
EGF (human recombinant) | 0.5 | Infinity | |
Other Components | |||
FBS | confidential | n/a | |
Heparin | 500.0 | Infinity | |
Hydrocortisone | 50.0 | Infinity |
Human Keratinocyte Growth Supplement (HKGS)
Cat No:
s0015
Human Melanocyte Growth Supplement (HMGS)
Cat No:
s0025
Microvascular Growth Supplement (MVGS)
Cat No:
s00525
Smooth Muscle Growth Supplement (SMGS)
Cat No:
s00725
Smooth Muscle Differentiation Supplement (SMDS)
Cat No:
s0085
Human Corneal Growth Supplement (HCGS)
Cat No:
s0095
EpiLife® Defined Growth Supplement (EDGS)
Cat No:
s0125
Mammary Epithelial Growth Supplement (MEGS)
Cat No:
s0155
S0165 - Human Melanocyte Growth Supplement-2, PMA-Free (HMGS-2)
Cat No:
s0165