Search Thermo Fisher Scientific
Catalog Number(s)
11054020Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
Glycine | 75.0 | 30.0 | 0.4 |
L-Arginine hydrochloride | 211.0 | 84.0 | 0.39810428 |
L-Cystine 2HCl | 313.0 | 63.0 | 0.20127796 |
L-Histidine hydrochloride-H2O | 210.0 | 42.0 | 0.2 |
L-Isoleucine | 131.0 | 105.0 | 0.8015267 |
L-Leucine | 131.0 | 105.0 | 0.8015267 |
L-Lysine hydrochloride | 183.0 | 146.0 | 0.7978142 |
L-Methionine | 149.0 | 30.0 | 0.20134228 |
L-Phenylalanine | 165.0 | 66.0 | 0.4 |
L-Serine | 105.0 | 42.0 | 0.4 |
L-Threonine | 119.0 | 95.0 | 0.79831934 |
L-Tryptophan | 204.0 | 16.0 | 0.078431375 |
L-Tyrosine disodium salt dihydrate | 261.0 | 104.0 | 0.39846742 |
L-Valine | 117.0 | 94.0 | 0.8034188 |
Vitamins | |||
Choline chloride | 140.0 | 4.0 | 0.028571429 |
D-Calcium pantothenate | 477.0 | 4.0 | 0.008385744 |
Folic Acid | 441.0 | 4.0 | 0.009070295 |
Niacinamide | 122.0 | 4.0 | 0.032786883 |
Pyridoxine hydrochloride | 206.0 | 4.0 | 0.019417476 |
Riboflavin | 376.0 | 0.4 | 0.0010638298 |
Thiamine hydrochloride | 337.0 | 4.0 | 0.011869436 |
i-Inositol | 180.0 | 7.2 | 0.04 |
Inorganic Salts | |||
Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 200.0 | 1.8018018 |
Ferric Nitrate (Fe(NO3)3"9H2O) | 404.0 | 0.1 | 2.4752476E-4 |
Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 97.67 | 0.8139166 |
Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
Sodium Bicarbonate (NaHCO3) | 84.0 | 3700.0 | 44.04762 |
Sodium Chloride (NaCl) | 58.0 | 6400.0 | 110.344826 |
Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 125.0 | 0.9057971 |
Other Components | |||
D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
Sodium Pyruvate | 110.0 | 110.0 | 1.0 |
1. Dulbecco, R. and Freeman, G. (1959) Virology 8:396.
11880 - DMEM, low glucose, pyruvate, no glutamine, no phenol red
Cat No:
11880036