Search Thermo Fisher Scientific
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
Glycine | 75.0 | 7.5 | 0.1 |
L-Alanine | 89.0 | 8.9 | 0.099999994 |
L-Arginine hydrochloride | 211.0 | 211.0 | 1.0 |
L-Asparagine-H2O | 150.0 | 15.01 | 0.10006667 |
L-Aspartic acid | 133.0 | 13.3 | 0.1 |
L-Cysteine hydrochloride-H2O | 176.0 | 35.12 | 0.19954544 |
L-Glutamic Acid | 147.0 | 14.7 | 0.1 |
L-Glutamine | 146.0 | 146.0 | 1.0 |
L-Histidine hydrochloride-H2O | 210.0 | 21.0 | 0.1 |
L-Isoleucine | 131.0 | 4.0 | 0.030534351 |
L-Leucine | 131.0 | 13.1 | 0.1 |
L-Lysine hydrochloride | 183.0 | 36.5 | 0.19945355 |
L-Methionine | 149.0 | 4.5 | 0.030201342 |
L-Phenylalanine | 165.0 | 5.0 | 0.030303031 |
L-Proline | 115.0 | 34.5 | 0.3 |
L-Serine | 105.0 | 10.5 | 0.1 |
L-Threonine | 119.0 | 11.9 | 0.099999994 |
L-Tryptophan | 204.0 | 2.04 | 0.01 |
L-Tyrosine disodium salt dihydrate | 262.0 | 7.81 | 0.02980916 |
L-Valine | 117.0 | 11.7 | 0.1 |
Vitamins | |||
Biotin | 244.0 | 0.0073 | 2.9918034E-5 |
Choline chloride | 140.0 | 14.0 | 0.1 |
D-Calcium pantothenate | 477.0 | 0.5 | 0.001048218 |
Folic Acid | 441.0 | 1.3 | 0.0029478457 |
Niacinamide | 122.0 | 0.036 | 2.9508196E-4 |
Pyridoxine hydrochloride | 206.0 | 0.06 | 2.9126214E-4 |
Riboflavin | 376.0 | 0.037 | 9.8404256E-5 |
Thiamine hydrochloride | 337.0 | 0.3 | 8.9020777E-4 |
Vitamin B12 | 1355.0 | 1.4 | 0.0010332103 |
i-Inositol | 180.0 | 18.0 | 0.1 |
Inorganic Salts | |||
Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 33.22 | 0.2992793 |
Cupric sulfate (CuSO4-5H2O) | 250.0 | 0.0025 | 1.0E-5 |
Ferric sulfate (FeSO4-7H2O) | 278.0 | 0.834 | 0.003 |
Magnesium Chloride (anhydrous) | 95.0 | 57.22 | 0.6023158 |
Potassium Chloride (KCl) | 75.0 | 223.6 | 2.9813335 |
Sodium Bicarbonate (NaHCO3) | 84.0 | 1176.0 | 14.0 |
Sodium Chloride (NaCl) | 58.0 | 7599.0 | 131.01724 |
Sodium Phosphate dibasic (Na2HPO4) anhydrous | 142.0 | 142.0 | 1.0 |
Zinc sulfate (ZnSO4-7H2O) | 288.0 | 0.863 | 0.0029965276 |
Other Components | |||
D-Glucose (Dextrose) | 180.0 | 1802.0 | 10.011111 |
Hypoxanthine Na | 159.0 | 4.77 | 0.03 |
Linoleic Acid | 280.0 | 0.084 | 2.9999999E-4 |
Lipoic Acid | 206.0 | 0.21 | 0.0010194174 |
Phenol Red | 376.4 | 1.2 | 0.0031880978 |
Putrescine 2HCl | 161.0 | 0.161 | 0.001 |
Sodium Pyruvate | 110.0 | 110.0 | 1.0 |
Thymidine | 242.0 | 0.7 | 0.002892562 |
1. Ham, R. G. (1965) Proc. Nat. Acad. Sci., 53, 288.
21765 - Ham's F-12 Nutrient Mix
Cat No:
21765029
,
21765037