Search Thermo Fisher Scientific
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
Glycine | 75.0 | 50.0 | 0.6666667 |
L-Alanine | 89.0 | 25.0 | 0.28089887 |
L-Arginine hydrochloride | 211.0 | 70.0 | 0.33175355 |
L-Aspartic acid | 133.0 | 30.0 | 0.22556391 |
L-Cysteine hydrochloride-H2O | 176.0 | 0.1 | 5.681818E-4 |
L-Cystine 2HCl | 240.0 | 26.0 | 0.108333334 |
L-Glutamic Acid | 147.0 | 75.0 | 0.5102041 |
L-Glutamine | 146.0 | 100.0 | 0.6849315 |
L-Histidine hydrochloride-H2O | 210.0 | 21.88 | 0.10419047 |
L-Hydroxyproline | 131.0 | 10.0 | 0.07633588 |
L-Isoleucine | 131.0 | 40.0 | 0.3053435 |
L-Leucine | 131.0 | 60.0 | 0.45801526 |
L-Lysine hydrochloride | 183.0 | 70.0 | 0.38251367 |
L-Methionine | 149.0 | 15.0 | 0.10067114 |
L-Phenylalanine | 165.0 | 25.0 | 0.15151516 |
L-Proline | 115.0 | 40.0 | 0.3478261 |
L-Serine | 105.0 | 25.0 | 0.23809524 |
L-Threonine | 119.0 | 30.0 | 0.25210086 |
L-Tryptophan | 204.0 | 10.0 | 0.04901961 |
L-Tyrosine disodium salt dihydrate | 261.0 | 58.0 | 0.22222222 |
L-Valine | 117.0 | 25.0 | 0.21367522 |
Vitamins | |||
Ascorbic Acid | 176.0 | 0.05 | 2.840909E-4 |
Biotin | 244.0 | 0.01 | 4.0983607E-5 |
Choline chloride | 140.0 | 0.5 | 0.0035714286 |
D-Calcium pantothenate | 477.0 | 0.01 | 2.096436E-5 |
Folic Acid | 441.0 | 0.01 | 2.2675737E-5 |
Menadione (Vitamin K3) | 172.0 | 0.01 | 5.8139532E-5 |
Niacinamide | 122.0 | 0.025 | 2.0491803E-4 |
Nicotinic acid (Niacin) | 123.0 | 0.025 | 2.0325204E-4 |
Para-Aminobenzoic Acid | 137.0 | 0.05 | 3.6496352E-4 |
Pyridoxal hydrochloride | 204.0 | 0.025 | 1.2254903E-4 |
Pyridoxine hydrochloride | 206.0 | 0.025 | 1.21359226E-4 |
Riboflavin | 376.0 | 0.01 | 2.6595744E-5 |
Thiamine hydrochloride | 337.0 | 0.01 | 2.967359E-5 |
Vitamin A (acetate) | 328.0 | 0.1 | 3.0487805E-4 |
Vitamin D2 (Calciferol) | 397.0 | 0.1 | 2.5188917E-4 |
alpha Tocopherol phos. Na salt | 554.7 | 0.01 | 1.8027762E-5 |
i-Inositol | 180.0 | 0.05 | 2.7777778E-4 |
Inorganic Salts | |||
Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 200.0 | 1.8018018 |
Ferric nitrate (Fe(NO3)-9H2O) | 404.0 | 0.7 | 0.0017326733 |
Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 97.67 | 0.8139166 |
Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
Sodium Bicarbonate (NaHCO3) | 84.0 | 2200.0 | 26.190475 |
Sodium Chloride (NaCl) | 58.0 | 6800.0 | 117.24138 |
Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 140.0 | 1.0144928 |
Other Components | |||
2-deoxy-D-ribose | 134.0 | 0.5 | 0.0037313432 |
Adenine sulfate | 404.0 | 10.0 | 0.024752475 |
Adenosine 5'-phosphate | 347.0 | 0.2 | 5.763689E-4 |
Adenosine 5'-triphosphate | 605.0 | 1.0 | 0.0016528926 |
Cholesterol | 387.0 | 0.2 | 5.1679584E-4 |
D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
Glutathione (reduced) | 307.0 | 0.05 | 1.6286645E-4 |
Guanine hydrochloride | 188.0 | 0.3 | 0.0015957447 |
Hypoxanthine Na | 136.0 | 0.4 | 0.0029411765 |
Phenol Red | 376.4 | 20.0 | 0.053134963 |
Ribose | 150.0 | 0.5 | 0.0033333334 |
Sodium Acetate | 82.0 | 50.0 | 0.6097561 |
Thymine | 126.0 | 0.3 | 0.0023809525 |
Tween 80® | 20.0 | Infinity | |
Uracil | 112.0 | 0.3 | 0.0026785715 |
Xanthine-Na | 152.0 | 0.34 | 0.0022368422 |
1. Morgan, J.F. and Campbell, M.E. (1955) J. Natl. Cancer Inst., 16:557.
2. Morgan, J.F., Morton, H.J. and Parker R.C. (1950) Proc. Soc. Exp. Biol. Med., 73:1.
31150 - Medium 199
Cat No:
31150022
,
31150030