Mouse kidney section. FluoCells® prepared slide #3.
FluoCells® prepared slide #3 (Cat. no. F24630) contains a mouse kidney section stained with a combination of fluorescent dyes. Alexa Fluor® 488 wheat germ agglutinin (Cat. no. W11261) is a green-fluorescent lectin that was used to label elements of the glomeruli and convoluted tubules. The filamentous actin prevalent in glomeruli and the brush border were stained with red-orange–fluorescent Alexa Fluor® 568 phalloidin (Cat. no. A12380). Finally, the nuclei were stained with the blue-fluorescent DNA stain DAPI (Cat. no. D1306, D3571, D21490). This pseudocolored image was acquired on a Zeiss confocal microscope located at the Institute of Neuroscience, University of Oregon.
FluoCells® prepared slide #3 (Cat. no. F24630) contains a mouse kidney section stained with a combination of fluorescent dyes. Alexa Fluor® 488 wheat germ agglutinin (Cat. no. W11261) is a green-fluorescent lectin that was used to label elements of the glomeruli and convoluted tubules. The filamentous actin prevalent in glomeruli and the brush border were stained with red-orange–fluorescent Alexa Fluor® 568 phalloidin (Cat. no. A12380). Finally, the nuclei were stained with the blue-fluorescent DNA stain DAPI (Cat. no. D1306, D3571, D21490).  This pseudocolored image was acquired on a Zeiss confocal microscope located at the Institute of Neuroscience, University of Oregon.
Related Products
Catalog # Name Size Price (USD) Qty
A12380 Alexa Fluor™ 568 Phalloidin, Orange-red, 578/600 nm Each
628.65
Online Exclusive
660.00
Save 31.35 (5%)
D1306 DAPI (4',6-diamidino-2-phenylindole, dihydrochloride), 10 mg, 10 mg Each
152.65
Online Exclusive
156.00
Save 3.35 (2%)
F24630 FluoCells™ Prepared Slide #3 (mouse kidney section with Alexa Fluor™ 488 WGA, Alexa Fluor™ 568 Phalloidin, and DAPI) Each
351.65
Online Exclusive
382.00
Save 30.35 (8%)
W11261 Wheat Germ Agglutinin, Alexa Fluor™ 488 Conjugate, 5 mg 5 mg
472.65
Online Exclusive
486.00
Save 13.35 (3%)
Related Images