Human metaphase chromosomes hybridized to centromere probes labeled with ChromaTide® OBEA-dCTP nucleotides.
Human metaphase chromosomes hybridized to centromere probes labeled with ChromaTide® OBEA-dCTP nucleotides. Probes specific to the a-satellite sequences from human chromosome 17, chromosome 15 and chromosome 1 were labeled using nick translation with ChromaTide® Alexa Fluor® 488-7-OBEA-dCTP (green; Cat. no. C21555), ChromaTide® Alexa Fluor® 594-7-OBEA-dCTP (red; Cat. no. C21558) and ChromaTide® Alexa Fluor® 647-12-OBEA-dCTP (pink; Cat. no. C21559), respectively. The probes were hybridized to metaphase spreads from peripheral blood lymphocyte cultures. The chromosomes were counterstained with Hoechst 33342 (blue) (Cat. no. H1399, H3570, H21492).
Human metaphase chromosomes hybridized to centromere probes labeled with ChromaTide® OBEA-dCTP nucleotides. Probes specific to the a-satellite sequences from human chromosome 17, chromosome 15 and chromosome 1 were labeled using nick translation with ChromaTide® Alexa Fluor® 488-7-OBEA-dCTP (green; Cat. no. C21555), ChromaTide® Alexa Fluor® 594-7-OBEA-dCTP (red; Cat. no. C21558) and ChromaTide® Alexa Fluor® 647-12-OBEA-dCTP (pink; Cat. no. C21559), respectively. The probes were hybridized to metaphase spreads from peripheral blood lymphocyte cultures. The chromosomes were counterstained with Hoechst 33342 (blue) (Cat. no. H1399, H3570, H21492).
Related Products
Catalog # Name Size Price (USD) Qty
H21492 Hoechst 33342, trihydrochloride trihydrate, FluoroPure™ grade, 100 mg, 100 mg Each
129.65
Online Exclusive
133.00
Save 3.35 (3%)
H3570 Hoechst 33342, trihydrochloride trihydrate, 10 mg/mL solution in water, 10 mL Each
142.65
Online Exclusive
147.00
Save 4.35 (3%)
Related Images