Endogenous alkaline phosphatase activity of osteoblast cells in a cartilaginous element of an adult zebrafish head.
Endogenous phosphatase activity of osteoblast cells in a cartilaginous element of an adult zebrafish head was localized using the ELF 97 Endogenous Phosphatase Detection Kit (E6601) to stain a cryosection. In addition to the yellow-green fluorescence of the ELF 97 alcohol precipitate, the section was stained with red-fluorescent Texas Red-X wheat germ agglutinin (W21405) and with the blue-fluorescent Hoechst 33342 nucleic acid stain (H1399, H3570, H21492). The triple-exposure image was acquired using bandpass filter sets appropriate for ELF 97 alcohol, Texas Red dye and DAPI.
Endogenous phosphatase activity of osteoblast cells in a cartilaginous element of an adult zebrafish head was localized using the ELF 97 Endogenous Phosphatase Detection Kit (E6601) to stain a cryosection. In addition to the yellow-green fluorescence of the ELF 97 alcohol precipitate, the section was stained with red-fluorescent Texas Red-X wheat germ agglutinin (W21405) and with the blue-fluorescent Hoechst 33342 nucleic acid stain (H1399, H3570, H21492). The triple-exposure image was acquired using bandpass filter sets appropriate for ELF 97 alcohol, Texas Red dye and DAPI.
Related Products
Catalog # Name Size Price (USD) Qty
H1399 Hoechst 33342, trihydrochloride trihydrate, 100 mg, 100 mg Each
109.65
Online Exclusive
113.00
Save 3.35 (3%)
H21492 Hoechst 33342, trihydrochloride trihydrate, FluoroPure™ grade, 100 mg, 100 mg Each
129.65
Online Exclusive
133.00
Save 3.35 (3%)
H3570 Hoechst 33342, trihydrochloride trihydrate, 10 mg/mL solution in water, 10 mL Each
142.65
Online Exclusive
147.00
Save 4.35 (3%)
W21405 Wheat Germ Agglutinin, Texas Red™-X Conjugate, 1 mg 1 mg
227.65
Online Exclusive
234.00
Save 6.35 (3%)
Related Images