Find valuable information.
Optimize your experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your research needs.
View the relevant questions below:
Having problems with your experiment?  Visit our
Troubleshooting page
General
Total RNA samples should be free of genomic DNA and we recommend including a DNase treatment with the RNA purification method. Contaminating genomic DNA may be amplified along with the RNA, which can lead to inaccurate measurement of whole transcriptome expression. In addition, the contaminating genomic DNA could cause over-estimation of the RNA amount.​
We strongly recommend against the use of nucleic acid-based and those that contain glycogen carriers during RNA purification because many have been shown to produce cDNA products in first-strand synthesis reaction. Choose a purification method or commercially available kit that is appropriate for your sample amount. For limited cell numbers, choose purification methods that enable purification of total RNA preparations from small amounts.
We do not avoid globin through primer design for the GeneChip microarrays. All our assays label and/or amplify globin RNA, and the amplified globins hybridize and generate array signals.
However, our WT (whole transcript) assays generate DNA as hybridization targets, which allow for the generation of strong signals of target transcripts even in the presence of globins. This is in contrast with the IVT (in vitro transcription) assays, where the binding of the generated labeled RNA targets and the array signals are weaker, due to globin interference. Therefore, it is necessary to run a globin reduction when using the IVT assays.
Yes, high amounts of ribosomal RNA (rRNA) can influence the amplification process. Ribosomal RNAs are the most abundant types of RNA present in cells, and they can compete with messenger RNAs (mRNAs), which are usually the targets that bind to the probes on the array, during the reverse transcription and amplification steps. This competition can lead to less efficient amplification of the mRNAs, which can impact the overall quality and reliability of the outcome data.
However, it's important to note that this doesn't necessarily cause "random" primer amplification.
The primers used in these steps are designed to be specific for the mRNAs depending on the assay used and also on the probes on the array, so the presence of excess rRNA shouldn't cause them to bind and amplify random sequences. But it can reduce the efficiency with which they amplify the specific sequences for which the probes are designed for.
As a best practice, depending on the nature of the input, we often recommend that you deplete rRNA from the samples before performing these steps, especially when working with total RNA samples. There are several commercially available kits and methods for rRNA depletion.
Yes, the GeneChip microarrays can use either poly(A)+ mRNA or total RNA as input material. The protocols provided in the manual describe procedures for preparing biotinylated target RNA from both purified eukaryotic poly(A)+ mRNA and total RNA samples. Good-quality mRNA has been successfully isolated from mammalian cells and tissues using specific kits, and this mRNA can be used as a template for cDNA synthesis. However, it is important to note that results obtained from samples prepared using different methods (total RNA vs poly(A)+ mRNA) may not be identical, so it is recommended to only compare samples prepared using the same sample preparation protocol.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
All human Axiom arrays (except the Axiom Genome-Wide Pan-African Array Set) require a total of 100 ng.
The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng or 100 ng per array.
Diploid plants and animals require 150 ng per array and polyploid plants and animals require 200 ng per array.
For Axiom Microbiome Arrays, a total of 50 ng of gDNA or 17.5 µL of cDNA reaction + 2.5 µL reduced TE buffer starting material is required per array.
Please refer to the Axiom 2.0 gDNA Sample Preparation Quick Reference for more details. Additionally, you can refer to pages 12-13 of the Axiom 2.0 Assay 96-Array Format Manual Workflow User Guide.
Quality recommendations:
- Starting gDNA must be double-stranded to support accurate concentration determination.
- gDNA must be of high purity.
- DNA should be free of DNA polymerase inhibitors. Examples of inhibitors include high concentrations of heme (from blood) and high concentrations of chelating agents (i.e., EDTA).
- The gDNA extraction/purification method should render DNA that is generally salt-free because high concentrations of particular salts can also inhibit enzyme reactions.
- DNA purity is indicated by OD260/OD280 and OD260/OD230 ratios. The OD260/OD280 ratio should be between 1.8 and 2.0 and the OD260/OD230 ratio should be greater than 1.5.
- DNA must not be degraded.
The approximate average size of gDNA may be assessed on a 1% agarose gel using an appropriate size standard control.
Approximately 90% of the DNA must be greater than 10 Kb in size. Control DNA can be run on the same gel for side-by-side comparison.
We recommend that DNA samples that do not meet these criteria be cleaned up as described under Genomic DNA Cleanup in the Axiom 2.0 Assay 96-Array Format Manual Workflow User Guide, pages 15-17.
gDNA extracted from stool samples has been tested for use with the Axiom Microbiome Array. DNA derived from FFPE (formalin-fixed paraffin-embedded) blocks should not be used with this assay.
Any extraction and purification kit that meets the starting gDNA requirements is acceptable. However, methods that require boiling or strong denaturants should not be used, as the DNA will be rendered single-stranded.
50 ng of gDNA (20 µL at 2.5 ng/µL) is optimal. The gDNA must also be double-stranded, of high purity (OD260/OD280 ratio of 1.8-2.0 and OD260/OD230 ratio greater than 1.5), and not degraded (90% of gDNA greater than 10 kb).
The tested sources of human gDNA used in the CytoScan Assay are blood and cell line samples.
The genomic DNA (gDNA) sample must be double-stranded, not degraded and free of any contaminants (e.g., PCR inhibitors and other human/non-human gDNA). The recommended starting amount is 250 ng (5 µL with 50 ng/µL) dissolved in low EDTA TE buffer. High EDTA concentration may negatively impact the downstream enzymatic reactions. We recommend running the gDNA samples on a 0.8-1% agarose gel for side-by side comparison with a control DNA (included in the kit). High-quality genomic DNA will run as a major band at approximately 10-20 kb on the gel.
For more detailed information please refer to the CytoScan Assay User Guide, page 9, Genomic DNA general requirements and recommendations section .
Gene expression
All catalog GeneChip™ expression arrays are designed to have a minimum sensitivity of 1:100,000. This concentration ratio corresponds roughly to a few copies of transcript per cell, or an approximate 1.5 pM concentration.
It is an optional step to collect the hybridization cocktail after the first overnight hybridization (16 hrs at 45 degrees C). The hyb cocktail can be collected from the array and transferred to a fresh microcentrifuge tube or well of a 96-well plate to save it. This can be used to re-hybridize new arrays, provided that there is no RNase contamination. The hyb cocktail transfer has to be stored on ice during the procedure and at -20 degrees C for long term storage. For more information please refer to the GeneChip WT PLUS Reagent Kit User Guide, Page 34 in Wash and stain the cartridge arrays section.
After hybridization, the GeneChip expression arrays can be stored in Wash Buffer A at 4 degrees C for up to 3 hours before proceeding with washing and staining. After washing and staining, the GeneChip expression arrays can be stored up to 16 hours in array holding buffer at 4 degrees C and in the dark before scanning. It is necessary to equilibrate arrays to room temperature before washing, staining, or scanning.
β-actin and GAPDH are used to assess RNA sample and assay quality by looking at the 3'/5' ratio. The 3'/5' ratio refers to the signal values of the 3' probe sets for actin and GAPDH being compared to the signal values of the corresponding 5' probe sets. The ratio is generally no more than 3 for the 1-cycle assay. Since the Affymetrix eukaryotic expression assay has an inherent 3' bias, a high 3' to 5' ratio may indicate degraded RNA or inefficient transcription of ds cDNA or biotinylated cRNA.
The 2-cycle assay typically gives higher 3' to 5' ratios than the 1-cycle assay, due to the additional cycle of amplification. There are occasions when the 3' to 5' ratio of one internal control gene is normal, but the 3' to 5' ratio of another internal control gene is high. This discrepancy in 3' to 5' ratios is most likely due to a specific transcript-related or image artifact issue and is not an indication of overall sample and assay quality.
The four transcripts are added to the hybridization cocktail at staggered concentrations. At 1.5 pM, bioB is at the detection limit for most expression arrays and is anticipated to be called Present at least 50% of the time. In contrast, the other controls should be called Present all of the time, with increasing Signal values (bioC, bioD, and cre, respectively).
For Research Use Only. Not for use in diagnostic procedures.